Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 117824895-117825152 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX18
Entrez Gene ID
8886
Gene Name
DEAD-box helicase 18
Gene Aliases
MrDb
UniGene
Hs.744922
Gene Chromosome Location
Chr.2: 117814679-117832377 on Build GRCh38
Transcripts
NM_006773.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
268993 c.1237T>A p.F413I Chr.2: 117824970-117824970 on Build GRCh38
239541 c.1354G>A p.V452I Chr.2: 117825087-117825087 on Build GRCh38
3565661 c.1361C>T p.A454V Chr.2: 117825094-117825094 on Build GRCh38
1660176 c.1355delT p.V452fs*38 Chr.2: 117825088-117825088 on Build GRCh38
418601 c.1243C>T p.L415L Chr.2: 117824976-117824976 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147447595 Chr.2: 117825068-117825068 on Build GRCh38
rs760534940 Chr.2: 117825048-117825048 on Build GRCh38
rs757130872 Chr.2: 117825045-117825045 on Build GRCh38
rs763636439 Chr.2: 117825105-117825105 on Build GRCh38
rs200592675 Chr.2: 117825004-117825004 on Build GRCh38
rs185105293 Chr.2: 117825074-117825074 on Build GRCh38
rs753544982 Chr.2: 117825107-117825107 on Build GRCh38
rs187619593 Chr.2: 117824931-117824931 on Build GRCh38
rs768907886 Chr.2: 117824961-117824961 on Build GRCh38
rs971794094 Chr.2: 117825021-117825021 on Build GRCh38
rs762223882 Chr.2: 117824999-117824999 on Build GRCh38
rs61732821 Chr.2: 117825035-117825035 on Build GRCh38
rs773402669 Chr.2: 117825090-117825090 on Build GRCh38
rs753524650 Chr.2: 117825041-117825041 on Build GRCh38
rs930406640 Chr.2: 117825039-117825039 on Build GRCh38
rs141313148 Chr.2: 117825107-117825109 on Build GRCh38
rs781406596 Chr.2: 117825083-117825083 on Build GRCh38
rs749132970 Chr.2: 117824924-117824924 on Build GRCh38
rs747160399 Chr.2: 117825073-117825073 on Build GRCh38
rs771378291 Chr.2: 117825094-117825094 on Build GRCh38
rs141912599 Chr.2: 117825038-117825038 on Build GRCh38
rs192548183 Chr.2: 117825015-117825015 on Build GRCh38
rs763357437 Chr.2: 117825091-117825091 on Build GRCh38
rs1022769630 Chr.2: 117825027-117825027 on Build GRCh38
rs765100115 Chr.2: 117825117-117825117 on Build GRCh38
rs767078619 Chr.2: 117825029-117825029 on Build GRCh38
rs528961378 Chr.2: 117825114-117825114 on Build GRCh38
rs757110375 Chr.2: 117824928-117824928 on Build GRCh38
rs745734977 Chr.2: 117824936-117824936 on Build GRCh38
rs866830200 Chr.2: 117825084-117825084 on Build GRCh38
rs375317000 Chr.2: 117824930-117824930 on Build GRCh38
rs774992319 Chr.2: 117825098-117825098 on Build GRCh38
rs545681587 Chr.2: 117824987-117824987 on Build GRCh38
rs575230561 Chr.2: 117825113-117825113 on Build GRCh38
rs1032380305 Chr.2: 117825081-117825081 on Build GRCh38
rs780011548 Chr.2: 117825070-117825070 on Build GRCh38
rs775390970 Chr.2: 117824955-117824955 on Build GRCh38
rs748307224 Chr.2: 117825086-117825086 on Build GRCh38
rs750232828 Chr.2: 117825119-117825119 on Build GRCh38
rs770229868 Chr.2: 117825087-117825087 on Build GRCh38
rs267598848 Chr.2: 117824972-117824972 on Build GRCh38
rs747252412 Chr.2: 117824956-117824956 on Build GRCh38
rs772196321 Chr.2: 117824942-117824942 on Build GRCh38
rs140766940 Chr.2: 117825059-117825059 on Build GRCh38
rs371410164 Chr.2: 117825019-117825019 on Build GRCh38
rs760315453 Chr.2: 117825102-117825102 on Build GRCh38
rs780431314 Chr.2: 117825080-117825080 on Build GRCh38

View Full Product Details