Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 159854608-159854845 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf204
Entrez Gene ID
284677
Gene Name
chromosome 1 open reading frame 204
Gene Aliases
-
UniGene
Hs.647718
Gene Chromosome Location
Chr.1: 159834474-159855347 on Build GRCh38
Transcripts
NM_001134233.1 NM_001013661.1

Gene Symbol
VSIG8
Entrez Gene ID
391123
Gene Name
V-set and immunoglobulin domain containing 8
Gene Aliases
-
UniGene
Hs.647718
Gene Chromosome Location
Chr.1: 159854316-159862657 on Build GRCh38
Transcripts
NM_001134233.1 NM_001013661.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6397933 c.305C>T p.P102L Chr.1: 159854784-159854784 on Build GRCh38
6533971 c.322G>C p.G108R Chr.1: 159854767-159854767 on Build GRCh38
6397932 c.1214C>T p.P405L Chr.1: 159854784-159854784 on Build GRCh38
6580403 c.1192C>A p.P398T Chr.1: 159854806-159854806 on Build GRCh38
6580405 c.283C>A p.P95T Chr.1: 159854806-159854806 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1050792398 Chr.1: 159854689-159854689 on Build GRCh38
rs1010822970 Chr.1: 159854798-159854798 on Build GRCh38
rs1039982032 Chr.1: 159854680-159854680 on Build GRCh38
rs565407941 Chr.1: 159854707-159854707 on Build GRCh38
rs115484427 Chr.1: 159854633-159854633 on Build GRCh38
rs751298673 Chr.1: 159854754-159854754 on Build GRCh38
rs999593539 Chr.1: 159854729-159854729 on Build GRCh38
rs936816781 Chr.1: 159854827-159854827 on Build GRCh38
rs573516875 Chr.1: 159854644-159854644 on Build GRCh38
rs1024313153 Chr.1: 159854819-159854819 on Build GRCh38
rs901519551 Chr.1: 159854692-159854692 on Build GRCh38
rs540814104 Chr.1: 159854659-159854659 on Build GRCh38
rs544815924 Chr.1: 159854751-159854751 on Build GRCh38
rs903022692 Chr.1: 159854716-159854716 on Build GRCh38
rs752424564 Chr.1: 159854810-159854810 on Build GRCh38
rs920288577 Chr.1: 159854667-159854667 on Build GRCh38
rs892743014 Chr.1: 159854666-159854666 on Build GRCh38
rs748108966 Chr.1: 159854805-159854805 on Build GRCh38
rs930269612 Chr.1: 159854685-159854685 on Build GRCh38
rs755583013 Chr.1: 159854813-159854813 on Build GRCh38
rs754868464 Chr.1: 159854767-159854767 on Build GRCh38
rs780923481 Chr.1: 159854786-159854786 on Build GRCh38
rs962019804 Chr.1: 159854768-159854768 on Build GRCh38
rs945713332 Chr.1: 159854669-159854669 on Build GRCh38
rs1024944039 Chr.1: 159854789-159854789 on Build GRCh38
rs1053104079 Chr.1: 159854655-159854655 on Build GRCh38
rs749346915 Chr.1: 159854700-159854700 on Build GRCh38
rs971790690 Chr.1: 159854658-159854658 on Build GRCh38
rs887593038 Chr.1: 159854708-159854708 on Build GRCh38
rs893826911 Chr.1: 159854794-159854794 on Build GRCh38
rs773099233 Chr.1: 159854766-159854766 on Build GRCh38
rs980856970 Chr.1: 159854638-159854638 on Build GRCh38
rs992086439 Chr.1: 159854776-159854776 on Build GRCh38
rs928113201 Chr.1: 159854637-159854637 on Build GRCh38
rs910430883 Chr.1: 159854690-159854690 on Build GRCh38
rs936869607 Chr.1: 159854651-159854651 on Build GRCh38

View Full Product Details