Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 12716170-12716427 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AADACL3
Entrez Gene ID
126767
Gene Name
arylacetamide deacetylase like 3
Gene Aliases
-
UniGene
Hs.464705
Gene Chromosome Location
Chr.1: 12716115-12728759 on Build GRCh38
Transcripts
NM_001103170.2 NR_111984.1

SNP Detail

SNP ID Chromosome Location
rs767486870 Chr.1: 12716335-12716335 on Build GRCh38
rs370828009 Chr.1: 12716290-12716290 on Build GRCh38
rs989820450 Chr.1: 12716283-12716283 on Build GRCh38
rs774648022 Chr.1: 12716317-12716317 on Build GRCh38
rs75374747 Chr.1: 12716340-12716340 on Build GRCh38
rs3000859 Chr.1: 12716341-12716341 on Build GRCh38
rs953059133 Chr.1: 12716391-12716391 on Build GRCh38
rs758029667 Chr.1: 12716367-12716367 on Build GRCh38
rs924613625 Chr.1: 12716233-12716233 on Build GRCh38
rs999061516 Chr.1: 12716284-12716284 on Build GRCh38
rs758064050 Chr.1: 12716200-12716200 on Build GRCh38
rs867514872 Chr.1: 12716192-12716192 on Build GRCh38
rs749414451 Chr.1: 12716309-12716309 on Build GRCh38
rs977482821 Chr.1: 12716229-12716229 on Build GRCh38
rs766392999 Chr.1: 12716349-12716349 on Build GRCh38
rs1012944434 Chr.1: 12716216-12716216 on Build GRCh38
rs768569348 Chr.1: 12716312-12716312 on Build GRCh38
rs917368863 Chr.1: 12716213-12716213 on Build GRCh38
rs374527992 Chr.1: 12716378-12716378 on Build GRCh38
rs3000860 Chr.1: 12716215-12716215 on Build GRCh38
rs781584453 Chr.1: 12716370-12716370 on Build GRCh38
rs552228879 Chr.1: 12716321-12716321 on Build GRCh38
rs756427903 Chr.1: 12716393-12716393 on Build GRCh38
rs199994410 Chr.1: 12716357-12716357 on Build GRCh38
rs541900788 Chr.1: 12716267-12716267 on Build GRCh38
rs770263359 Chr.1: 12716293-12716293 on Build GRCh38
rs202247265 Chr.1: 12716365-12716365 on Build GRCh38
rs935222723 Chr.1: 12716266-12716266 on Build GRCh38
rs764582309 Chr.1: 12716360-12716360 on Build GRCh38
rs907345675 Chr.1: 12716274-12716274 on Build GRCh38
rs775586305 Chr.1: 12716303-12716303 on Build GRCh38
rs780631807 Chr.1: 12716394-12716394 on Build GRCh38
rs1049052532 Chr.1: 12716203-12716203 on Build GRCh38

View Full Product Details