Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150553586-150553847 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTSL4
Entrez Gene ID
54507
Gene Name
ADAMTS like 4
Gene Aliases
ADAMTSL-4, ECTOL2, TSRC1
UniGene
Hs.516243
Gene Chromosome Location
Chr.1: 150549369-150560937 on Build GRCh38
Transcripts
NM_001288607.1 NM_001288608.1 NM_019032.5 NM_025008.4 XM_011509644.2 XM_011509645.2 XM_011509648.2 XM_011509649.2 XM_011509650.2 XM_017001506.1

Gene Symbol
LOC100289061
Entrez Gene ID
100289061
Gene Name
uncharacterized LOC100289061
Gene Aliases
-
UniGene
Hs.677561
Gene Chromosome Location
Chr.1: 150548564-150556700 on Build GRCh38
Transcripts
NM_001288607.1 NM_001288608.1 NM_019032.5 NM_025008.4 XM_011509644.2 XM_011509645.2 XM_011509648.2 XM_011509649.2 XM_011509650.2 XM_017001506.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6594590 c.808G>A p.G270R Chr.1: 150553799-150553799 on Build GRCh38
5010602 c.653A>G p.E218G Chr.1: 150553644-150553644 on Build GRCh38
5682934 c.816T>A p.G272G Chr.1: 150553807-150553807 on Build GRCh38
5353109 c.797C>T p.T266M Chr.1: 150553788-150553788 on Build GRCh38
1333837 c.798G>A p.T266T Chr.1: 150553789-150553789 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs587708719 Chr.1: 150553612-150553612 on Build GRCh38
rs769200929 Chr.1: 150553830-150553830 on Build GRCh38
rs794726688 Chr.1: 150553817-150553827 on Build GRCh38
rs765546357 Chr.1: 150553819-150553819 on Build GRCh38
rs377120955 Chr.1: 150553755-150553755 on Build GRCh38
rs145240194 Chr.1: 150553665-150553665 on Build GRCh38
rs763906182 Chr.1: 150553807-150553807 on Build GRCh38
rs758076428 Chr.1: 150553752-150553760 on Build GRCh38
rs764545096 Chr.1: 150553747-150553747 on Build GRCh38
rs756937513 Chr.1: 150553626-150553626 on Build GRCh38
rs761084716 Chr.1: 150553730-150553731 on Build GRCh38
rs587616373 Chr.1: 150553735-150553735 on Build GRCh38
rs746341961 Chr.1: 150553642-150553642 on Build GRCh38
rs779305942 Chr.1: 150553638-150553638 on Build GRCh38
rs777313892 Chr.1: 150553771-150553771 on Build GRCh38
rs768188442 Chr.1: 150553664-150553665 on Build GRCh38
rs368893787 Chr.1: 150553697-150553697 on Build GRCh38
rs587728833 Chr.1: 150553777-150553777 on Build GRCh38
rs753550085 Chr.1: 150553817-150553817 on Build GRCh38
rs779179264 Chr.1: 150553736-150553736 on Build GRCh38
rs587626697 Chr.1: 150553789-150553789 on Build GRCh38
rs749330047 Chr.1: 150553617-150553617 on Build GRCh38
rs150189250 Chr.1: 150553670-150553670 on Build GRCh38
rs377690308 Chr.1: 150553706-150553706 on Build GRCh38
rs115104834 Chr.1: 150553613-150553613 on Build GRCh38
rs771998925 Chr.1: 150553788-150553788 on Build GRCh38
rs41317517 Chr.1: 150553794-150553794 on Build GRCh38
rs762383955 Chr.1: 150553667-150553667 on Build GRCh38
rs775965947 Chr.1: 150553700-150553703 on Build GRCh38
rs898476873 Chr.1: 150553680-150553680 on Build GRCh38
rs772677738 Chr.1: 150553650-150553650 on Build GRCh38
rs1028528076 Chr.1: 150553708-150553708 on Build GRCh38
rs759041116 Chr.1: 150553785-150553785 on Build GRCh38
rs142905991 Chr.1: 150553745-150553745 on Build GRCh38
rs757300782 Chr.1: 150553631-150553631 on Build GRCh38
rs767427119 Chr.1: 150553701-150553701 on Build GRCh38
rs147803174 Chr.1: 150553658-150553658 on Build GRCh38
rs145673891 Chr.1: 150553740-150553740 on Build GRCh38
rs747366202 Chr.1: 150553656-150553656 on Build GRCh38
rs587698772 Chr.1: 150553606-150553606 on Build GRCh38
rs376319833 Chr.1: 150553818-150553818 on Build GRCh38
rs752465710 Chr.1: 150553607-150553607 on Build GRCh38
rs375539424 Chr.1: 150553669-150553669 on Build GRCh38
rs776049548 Chr.1: 150553652-150553652 on Build GRCh38
rs372604857 Chr.1: 150553734-150553734 on Build GRCh38
rs199705015 Chr.1: 150553726-150553726 on Build GRCh38
rs140535012 Chr.1: 150553739-150553739 on Build GRCh38
rs751125855 Chr.1: 150553806-150553807 on Build GRCh38
rs765800065 Chr.1: 150553774-150553775 on Build GRCh38
rs781580955 Chr.1: 150553653-150553653 on Build GRCh38
rs752551862 Chr.1: 150553727-150553727 on Build GRCh38
rs369322301 Chr.1: 150553779-150553779 on Build GRCh38

View Full Product Details