Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 35619885-35620116 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FJX1
Entrez Gene ID
24147
Gene Name
four jointed box 1
Gene Aliases
-
UniGene
Hs.39384
Gene Chromosome Location
Chr.11: 35618187-35620873 on Build GRCh38
Transcripts
NM_014344.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4843849 c.1251C>T p.L417L Chr.11: 35619887-35619887 on Build GRCh38
4032591 c.1294G>A p.G432S Chr.11: 35619930-35619930 on Build GRCh38
5361023 c.1278C>T p.H426H Chr.11: 35619914-35619914 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs898136944 Chr.11: 35620089-35620089 on Build GRCh38
rs545848130 Chr.11: 35620062-35620062 on Build GRCh38
rs936013183 Chr.11: 35619954-35619954 on Build GRCh38
rs768911636 Chr.11: 35619977-35619977 on Build GRCh38
rs946193193 Chr.11: 35619982-35619982 on Build GRCh38
rs1055440232 Chr.11: 35620054-35620054 on Build GRCh38
rs906915925 Chr.11: 35619998-35619998 on Build GRCh38
rs971650913 Chr.11: 35620029-35620029 on Build GRCh38
rs768233361 Chr.11: 35620015-35620015 on Build GRCh38
rs56272465 Chr.11: 35620004-35620004 on Build GRCh38
rs368400572 Chr.11: 35619988-35619988 on Build GRCh38
rs780119678 Chr.11: 35620025-35620026 on Build GRCh38
rs770875774 Chr.11: 35619947-35619947 on Build GRCh38
rs966536379 Chr.11: 35619932-35619932 on Build GRCh38
rs999517148 Chr.11: 35620049-35620049 on Build GRCh38
rs1013073600 Chr.11: 35620014-35620014 on Build GRCh38
rs769940544 Chr.11: 35620000-35620000 on Build GRCh38
rs1038461737 Chr.11: 35619970-35619970 on Build GRCh38
rs779030205 Chr.11: 35619958-35619958 on Build GRCh38
rs755737470 Chr.11: 35619920-35619920 on Build GRCh38
rs777561243 Chr.11: 35619930-35619930 on Build GRCh38
rs564019032 Chr.11: 35619940-35619940 on Build GRCh38
rs747078192 Chr.11: 35619976-35619976 on Build GRCh38
rs777076057 Chr.11: 35619967-35619968 on Build GRCh38
rs913518318 Chr.11: 35619968-35619968 on Build GRCh38
rs957030615 Chr.11: 35620068-35620068 on Build GRCh38

View Full Product Details