Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89651422-89651683 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHMP1A
Entrez Gene ID
5119
Gene Name
charged multivesicular body protein 1A
Gene Aliases
CHMP1, PCH8, PCOLN3, PRSM1, VPS46-1, VPS46A
UniGene
Hs.589427
Gene Chromosome Location
Chr.16: 89644431-89657785 on Build GRCh38
Transcripts
NM_001083314.3 NM_002768.4 NR_046418.2 XM_011523098.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4846823 c.16G>C p.E6Q Chr.16: 89651638-89651638 on Build GRCh38
5470604 c.84G>A p.A28A Chr.16: 89651590-89651590 on Build GRCh38
6299776 c.71G>A p.G24D Chr.16: 89651583-89651583 on Build GRCh38
4846824 c.36G>C p.A12A Chr.16: 89651638-89651638 on Build GRCh38
5363714 c.11A>G p.H4R Chr.16: 89651643-89651643 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201732385 Chr.16: 89651565-89651565 on Build GRCh38
rs750079319 Chr.16: 89651657-89651657 on Build GRCh38
rs770111037 Chr.16: 89651611-89651611 on Build GRCh38
rs984994926 Chr.16: 89651603-89651603 on Build GRCh38
rs8052569 Chr.16: 89651464-89651464 on Build GRCh38
rs397515426 Chr.16: 89651586-89651586 on Build GRCh38
rs367804633 Chr.16: 89651642-89651642 on Build GRCh38
rs986506515 Chr.16: 89651639-89651639 on Build GRCh38
rs371305175 Chr.16: 89651593-89651593 on Build GRCh38
rs763310272 Chr.16: 89651616-89651616 on Build GRCh38
rs191795485 Chr.16: 89651451-89651451 on Build GRCh38
rs763644107 Chr.16: 89651559-89651559 on Build GRCh38
rs772440021 Chr.16: 89651463-89651463 on Build GRCh38
rs200710259 Chr.16: 89651641-89651641 on Build GRCh38
rs1035352365 Chr.16: 89651498-89651498 on Build GRCh38
rs374315039 Chr.16: 89651613-89651613 on Build GRCh38
rs183210691 Chr.16: 89651608-89651608 on Build GRCh38
rs778561341 Chr.16: 89651578-89651578 on Build GRCh38
rs398122918 Chr.16: 89651659-89651659 on Build GRCh38
rs187184327 Chr.16: 89651623-89651623 on Build GRCh38
rs890881071 Chr.16: 89651513-89651513 on Build GRCh38
rs919633068 Chr.16: 89651461-89651461 on Build GRCh38
rs760130264 Chr.16: 89651558-89651558 on Build GRCh38
rs763354032 Chr.16: 89651551-89651551 on Build GRCh38
rs748373933 Chr.16: 89651609-89651609 on Build GRCh38
rs866256126 Chr.16: 89651583-89651583 on Build GRCh38
rs753406733 Chr.16: 89651652-89651652 on Build GRCh38
rs200605916 Chr.16: 89651660-89651660 on Build GRCh38
rs773691319 Chr.16: 89651538-89651538 on Build GRCh38
rs753459727 Chr.16: 89651560-89651560 on Build GRCh38
rs888996270 Chr.16: 89651460-89651460 on Build GRCh38
rs973980395 Chr.16: 89651456-89651456 on Build GRCh38
rs749669179 Chr.16: 89651602-89651604 on Build GRCh38
rs764880753 Chr.16: 89651653-89651653 on Build GRCh38
rs762172057 Chr.16: 89651528-89651528 on Build GRCh38
rs998548907 Chr.16: 89651476-89651476 on Build GRCh38
rs376691010 Chr.16: 89651590-89651590 on Build GRCh38
rs761485854 Chr.16: 89651663-89651663 on Build GRCh38
rs752012419 Chr.16: 89651555-89651555 on Build GRCh38
rs1042896583 Chr.16: 89651651-89651651 on Build GRCh38
rs776940457 Chr.16: 89651522-89651522 on Build GRCh38
rs766851415 Chr.16: 89651553-89651553 on Build GRCh38
rs142060347 Chr.16: 89651604-89651604 on Build GRCh38
rs780005367 Chr.16: 89651591-89651591 on Build GRCh38
rs765484837 Chr.16: 89651537-89651537 on Build GRCh38

View Full Product Details