Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 135936852-135937081 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL21
Entrez Gene ID
26223
Gene Name
F-box and leucine rich repeat protein 21 (gene/pseudogene)
Gene Aliases
FBL3B, FBXL3B, FBXL3P, Fbl21
UniGene
Hs.591275
Gene Chromosome Location
Chr.5: 135930317-135941678 on Build GRCh38
Transcripts
NM_012159.4 NR_132347.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
228068 c.284C>T p.S95L Chr.5: 135936878-135936878 on Build GRCh38
1582930 c.303T>A p.H101Q Chr.5: 135936897-135936897 on Build GRCh38
6700372 c.260T>G p.F87C Chr.5: 135936854-135936854 on Build GRCh38
4809061 c.337T>C p.F113L Chr.5: 135936931-135936931 on Build GRCh38
4779629 c.354T>C p.Y118Y Chr.5: 135936948-135936948 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777227198 Chr.5: 135936894-135936894 on Build GRCh38
rs747709719 Chr.5: 135936891-135936891 on Build GRCh38
rs777006081 Chr.5: 135937011-135937011 on Build GRCh38
rs570504337 Chr.5: 135936898-135936898 on Build GRCh38
rs768925285 Chr.5: 135936974-135936974 on Build GRCh38
rs567837954 Chr.5: 135937050-135937050 on Build GRCh38
rs760536408 Chr.5: 135936978-135936978 on Build GRCh38
rs373362472 Chr.5: 135937021-135937021 on Build GRCh38
rs958043343 Chr.5: 135936940-135936940 on Build GRCh38
rs769094789 Chr.5: 135936920-135936920 on Build GRCh38
rs753693302 Chr.5: 135936992-135936992 on Build GRCh38
rs774242974 Chr.5: 135936938-135936938 on Build GRCh38
rs963928192 Chr.5: 135937001-135937001 on Build GRCh38
rs372673285 Chr.5: 135936899-135936899 on Build GRCh38
rs768879045 Chr.5: 135936935-135936935 on Build GRCh38
rs761980960 Chr.5: 135936945-135936945 on Build GRCh38
rs778651649 Chr.5: 135937004-135937004 on Build GRCh38
rs150784167 Chr.5: 135937003-135937003 on Build GRCh38
rs116801846 Chr.5: 135936988-135936988 on Build GRCh38
rs767322265 Chr.5: 135936964-135936964 on Build GRCh38
rs763094669 Chr.5: 135936930-135936930 on Build GRCh38
rs749608542 Chr.5: 135936881-135936881 on Build GRCh38
rs567597686 Chr.5: 135936888-135936888 on Build GRCh38
rs775684252 Chr.5: 135936927-135936927 on Build GRCh38
rs988139150 Chr.5: 135936982-135936982 on Build GRCh38
rs369084967 Chr.5: 135936890-135936890 on Build GRCh38
rs564399844 Chr.5: 135936917-135936917 on Build GRCh38
rs976866365 Chr.5: 135937027-135937027 on Build GRCh38
rs999308205 Chr.5: 135936937-135936937 on Build GRCh38
rs771294420 Chr.5: 135936893-135936893 on Build GRCh38
rs750420386 Chr.5: 135936977-135936977 on Build GRCh38
rs376929937 Chr.5: 135936881-135936881 on Build GRCh38

View Full Product Details