Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 30165775-30166042 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN17
Entrez Gene ID
26285
Gene Name
claudin 17
Gene Aliases
-
UniGene
Hs.258589
Gene Chromosome Location
Chr.21: 30165923-30166653 on Build GRCh38
Transcripts
NM_012131.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5726278 c.580G>T p.G194W Chr.21: 30166038-30166038 on Build GRCh38
1615961 c.600T>A p.P200P Chr.21: 30166018-30166018 on Build GRCh38
6161036 c.670G>T p.V224F Chr.21: 30165948-30165948 on Build GRCh38
1029846 c.607C>T p.R203C Chr.21: 30166011-30166011 on Build GRCh38
1413585 c.579A>G p.Q193Q Chr.21: 30166039-30166039 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs188114857 Chr.21: 30165959-30165959 on Build GRCh38
rs755362951 Chr.21: 30166018-30166018 on Build GRCh38
rs890509905 Chr.21: 30165897-30165897 on Build GRCh38
rs770759302 Chr.21: 30165994-30165994 on Build GRCh38
rs563005963 Chr.21: 30165936-30165936 on Build GRCh38
rs182327550 Chr.21: 30166020-30166020 on Build GRCh38
rs767126875 Chr.21: 30165924-30165924 on Build GRCh38
rs753480841 Chr.21: 30165940-30165940 on Build GRCh38
rs373695346 Chr.21: 30165833-30165833 on Build GRCh38
rs756233700 Chr.21: 30165898-30165899 on Build GRCh38
rs1012807011 Chr.21: 30165978-30165978 on Build GRCh38
rs148953477 Chr.21: 30165827-30165827 on Build GRCh38
rs774403452 Chr.21: 30165913-30165913 on Build GRCh38
rs770683030 Chr.21: 30165893-30165893 on Build GRCh38
rs574128183 Chr.21: 30165921-30165921 on Build GRCh38
rs143848087 Chr.21: 30166011-30166011 on Build GRCh38
rs576177657 Chr.21: 30166012-30166012 on Build GRCh38
rs57014707 Chr.21: 30165835-30165835 on Build GRCh38
rs772069765 Chr.21: 30166001-30166001 on Build GRCh38
rs377679902 Chr.21: 30165990-30165990 on Build GRCh38
rs528102692 Chr.21: 30165886-30165886 on Build GRCh38
rs143710208 Chr.21: 30165872-30165872 on Build GRCh38
rs759417826 Chr.21: 30165995-30165995 on Build GRCh38
rs183838947 Chr.21: 30165928-30165928 on Build GRCh38
rs935884228 Chr.21: 30165829-30165829 on Build GRCh38
rs563734261 Chr.21: 30166010-30166010 on Build GRCh38
rs778036499 Chr.21: 30165989-30165989 on Build GRCh38
rs780091585 Chr.21: 30165901-30165905 on Build GRCh38
rs781665299 Chr.21: 30165980-30165980 on Build GRCh38
rs749537388 Chr.21: 30165986-30165987 on Build GRCh38
rs1027901826 Chr.21: 30165979-30165979 on Build GRCh38
rs138251734 Chr.21: 30165982-30165982 on Build GRCh38
rs763764272 Chr.21: 30165939-30165939 on Build GRCh38
rs775601768 Chr.21: 30166006-30166006 on Build GRCh38
rs545915811 Chr.21: 30165826-30165826 on Build GRCh38
rs752048768 Chr.21: 30165962-30165962 on Build GRCh38
rs748182900 Chr.21: 30165981-30165981 on Build GRCh38
rs146572430 Chr.21: 30165954-30165954 on Build GRCh38
rs755415911 Chr.21: 30165972-30165972 on Build GRCh38
rs776149659 Chr.21: 30165942-30165942 on Build GRCh38

View Full Product Details