Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 2393906-2394179 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD81
Entrez Gene ID
975
Gene Name
CD81 molecule
Gene Aliases
CVID6, S5.7, TAPA1, TSPAN28
UniGene
Hs.54457
Gene Chromosome Location
Chr.11: 2376177-2397419 on Build GRCh38
Transcripts
NM_001297649.1 NM_004356.3 XM_011520492.2 XM_017018598.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
687682 c.193C>G p.L65V Chr.11: 2394106-2394106 on Build GRCh38
3869000 c.229T>G p.F77V Chr.11: 2394142-2394142 on Build GRCh38
5372281 c.198C>T p.I66I Chr.11: 2394111-2394111 on Build GRCh38
687681 c.208G>A p.A70T Chr.11: 2394121-2394121 on Build GRCh38
234517 c.222C>T p.F74F Chr.11: 2394135-2394135 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750541732 Chr.11: 2394020-2394020 on Build GRCh38
rs758070892 Chr.11: 2394112-2394112 on Build GRCh38
rs753868647 Chr.11: 2394097-2394097 on Build GRCh38
rs975526745 Chr.11: 2393966-2393966 on Build GRCh38
rs115691463 Chr.11: 2393967-2393967 on Build GRCh38
rs751585256 Chr.11: 2393960-2393960 on Build GRCh38
rs776067002 Chr.11: 2394053-2394053 on Build GRCh38
rs912791937 Chr.11: 2393929-2393929 on Build GRCh38
rs200370609 Chr.11: 2394046-2394046 on Build GRCh38
rs779682894 Chr.11: 2393986-2393986 on Build GRCh38
rs566864289 Chr.11: 2394003-2394003 on Build GRCh38
rs754704039 Chr.11: 2394104-2394104 on Build GRCh38
rs759925248 Chr.11: 2393932-2393932 on Build GRCh38
rs749842476 Chr.11: 2394152-2394152 on Build GRCh38
rs751819789 Chr.11: 2394010-2394010 on Build GRCh38
rs769537594 Chr.11: 2393982-2393982 on Build GRCh38
rs752987997 Chr.11: 2394008-2394008 on Build GRCh38
rs375102275 Chr.11: 2394050-2394050 on Build GRCh38
rs748805462 Chr.11: 2394079-2394080 on Build GRCh38
rs1002124521 Chr.11: 2394024-2394024 on Build GRCh38
rs765723367 Chr.11: 2393935-2393935 on Build GRCh38
rs781049102 Chr.11: 2393973-2393973 on Build GRCh38
rs763439204 Chr.11: 2394009-2394009 on Build GRCh38
rs946751082 Chr.11: 2393934-2393934 on Build GRCh38
rs562261414 Chr.11: 2394090-2394090 on Build GRCh38
rs564371743 Chr.11: 2394108-2394108 on Build GRCh38
rs899976817 Chr.11: 2393951-2393951 on Build GRCh38
rs749033788 Chr.11: 2394028-2394028 on Build GRCh38
rs765890404 Chr.11: 2394006-2394006 on Build GRCh38
rs138186781 Chr.11: 2394036-2394036 on Build GRCh38
rs770534399 Chr.11: 2394126-2394126 on Build GRCh38
rs771673278 Chr.11: 2394040-2394040 on Build GRCh38
rs759831828 Chr.11: 2394113-2394113 on Build GRCh38
rs1989896 Chr.11: 2394062-2394062 on Build GRCh38
rs771416528 Chr.11: 2393995-2393995 on Build GRCh38
rs777575968 Chr.11: 2394120-2394120 on Build GRCh38
rs199682895 Chr.11: 2394089-2394089 on Build GRCh38
rs780098400 Chr.11: 2394026-2394026 on Build GRCh38
rs747983381 Chr.11: 2394111-2394111 on Build GRCh38
rs772822570 Chr.11: 2394041-2394041 on Build GRCh38
rs61731556 Chr.11: 2394153-2394153 on Build GRCh38
rs781280763 Chr.11: 2394019-2394019 on Build GRCh38
rs11546013 Chr.11: 2394147-2394147 on Build GRCh38
rs778112024 Chr.11: 2393959-2393959 on Build GRCh38
rs775840442 Chr.11: 2394135-2394135 on Build GRCh38
rs1044289756 Chr.11: 2393945-2393945 on Build GRCh38
rs756024872 Chr.11: 2394025-2394025 on Build GRCh38
rs757277562 Chr.11: 2394013-2394013 on Build GRCh38
rs558037994 Chr.11: 2394001-2394001 on Build GRCh38
rs763215788 Chr.11: 2394054-2394054 on Build GRCh38
rs749158217 Chr.11: 2393987-2393987 on Build GRCh38
rs758523706 Chr.11: 2393949-2393949 on Build GRCh38
rs777192545 Chr.11: 2394002-2394002 on Build GRCh38
rs768183072 Chr.11: 2393988-2393988 on Build GRCh38
rs746515946 Chr.11: 2394121-2394121 on Build GRCh38
rs745826699 Chr.11: 2393975-2393975 on Build GRCh38
rs574045896 Chr.11: 2394037-2394037 on Build GRCh38
rs774406198 Chr.11: 2394058-2394058 on Build GRCh38
rs764569621 Chr.11: 2394055-2394055 on Build GRCh38
rs768505089 Chr.11: 2394034-2394034 on Build GRCh38
rs753141911 Chr.11: 2393942-2393942 on Build GRCh38
rs533912152 Chr.11: 2394004-2394004 on Build GRCh38
rs575360607 Chr.11: 2394077-2394077 on Build GRCh38
rs769074665 Chr.11: 2394156-2394156 on Build GRCh38

View Full Product Details