Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 84617446-84617671 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COTL1
Entrez Gene ID
23406
Gene Name
coactosin like F-actin binding protein 1
Gene Aliases
CLP
UniGene
Hs.289092
Gene Chromosome Location
Chr.16: 84565596-84618096 on Build GRCh38
Transcripts
NM_021149.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
25209 c.147C>T p.I49I Chr.16: 84617514-84617514 on Build GRCh38
3818658 c.144C>G p.F48L Chr.16: 84617517-84617517 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1022414174 Chr.16: 84617607-84617608 on Build GRCh38
rs747764536 Chr.16: 84617592-84617592 on Build GRCh38
rs780723572 Chr.16: 84617526-84617526 on Build GRCh38
rs373858893 Chr.16: 84617627-84617627 on Build GRCh38
rs773589248 Chr.16: 84617624-84617624 on Build GRCh38
rs17855658 Chr.16: 84617548-84617548 on Build GRCh38
rs756346377 Chr.16: 84617560-84617560 on Build GRCh38
rs752989202 Chr.16: 84617513-84617513 on Build GRCh38
rs931116360 Chr.16: 84617647-84617647 on Build GRCh38
rs201009101 Chr.16: 84617552-84617552 on Build GRCh38
rs534225803 Chr.16: 84617576-84617576 on Build GRCh38
rs958141751 Chr.16: 84617509-84617509 on Build GRCh38
rs1044744516 Chr.16: 84617536-84617536 on Build GRCh38
rs759670407 Chr.16: 84617601-84617602 on Build GRCh38
rs370977924 Chr.16: 84617486-84617486 on Build GRCh38
rs767578234 Chr.16: 84617606-84617607 on Build GRCh38
rs759954602 Chr.16: 84617504-84617504 on Build GRCh38
rs751705777 Chr.16: 84617544-84617544 on Build GRCh38
rs772724957 Chr.16: 84617606-84617606 on Build GRCh38
rs1034995061 Chr.16: 84617591-84617591 on Build GRCh38
rs775626461 Chr.16: 84617465-84617465 on Build GRCh38
rs771472148 Chr.16: 84617593-84617593 on Build GRCh38
rs772738429 Chr.16: 84617503-84617503 on Build GRCh38
rs191019807 Chr.16: 84617638-84617638 on Build GRCh38
rs866624181 Chr.16: 84617497-84617497 on Build GRCh38
rs373998542 Chr.16: 84617491-84617491 on Build GRCh38
rs202135648 Chr.16: 84617547-84617547 on Build GRCh38
rs968662845 Chr.16: 84617616-84617616 on Build GRCh38
rs376359334 Chr.16: 84617476-84617476 on Build GRCh38
rs1018303998 Chr.16: 84617632-84617632 on Build GRCh38
rs369959700 Chr.16: 84617470-84617470 on Build GRCh38
rs562743599 Chr.16: 84617488-84617488 on Build GRCh38
rs913938964 Chr.16: 84617537-84617537 on Build GRCh38
rs764533632 Chr.16: 84617529-84617529 on Build GRCh38
rs140416122 Chr.16: 84617566-84617566 on Build GRCh38
rs770527264 Chr.16: 84617625-84617625 on Build GRCh38
rs879941912 Chr.16: 84617644-84617644 on Build GRCh38
rs781542652 Chr.16: 84617549-84617549 on Build GRCh38
rs774126238 Chr.16: 84617582-84617582 on Build GRCh38
rs199652382 Chr.16: 84617506-84617506 on Build GRCh38
rs113495648 Chr.16: 84617646-84617646 on Build GRCh38
rs1040111316 Chr.16: 84617475-84617475 on Build GRCh38
rs1012827226 Chr.16: 84617532-84617532 on Build GRCh38
rs758930676 Chr.16: 84617527-84617527 on Build GRCh38
rs768764476 Chr.16: 84617578-84617578 on Build GRCh38
rs549222468 Chr.16: 84617464-84617464 on Build GRCh38

View Full Product Details