Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 54816952-54817221 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LEXM
Entrez Gene ID
163747
Gene Name
lymphocyte expansion molecule
Gene Aliases
C1orf177, LEM
UniGene
Hs.376018
Gene Chromosome Location
Chr.1: 54806063-54842264 on Build GRCh38
Transcripts
NM_001110533.1 NM_152607.2 XM_017000480.1 XM_017000481.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6615195 c.1169G>A p.R390Q Chr.1: 54817107-54817107 on Build GRCh38
5847011 c.1121G>A p.R374Q Chr.1: 54817059-54817059 on Build GRCh38
5641205 c.1116A>G p.R372R Chr.1: 54817054-54817054 on Build GRCh38
4667348 c.1087C>A p.L363M Chr.1: 54817025-54817025 on Build GRCh38
4009000 c.1067G>A p.G356D Chr.1: 54817005-54817005 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs542343089 Chr.1: 54817005-54817005 on Build GRCh38
rs886213794 Chr.1: 54817070-54817070 on Build GRCh38
rs759516208 Chr.1: 54817168-54817168 on Build GRCh38
rs746394709 Chr.1: 54817054-54817054 on Build GRCh38
rs150463971 Chr.1: 54817048-54817048 on Build GRCh38
rs780615153 Chr.1: 54817141-54817141 on Build GRCh38
rs776598188 Chr.1: 54817090-54817090 on Build GRCh38
rs756422331 Chr.1: 54817133-54817133 on Build GRCh38
rs751194956 Chr.1: 54817164-54817164 on Build GRCh38
rs531676172 Chr.1: 54817004-54817004 on Build GRCh38
rs148287584 Chr.1: 54817065-54817065 on Build GRCh38
rs752503910 Chr.1: 54817111-54817111 on Build GRCh38
rs373250023 Chr.1: 54816983-54816983 on Build GRCh38
rs1016315254 Chr.1: 54816978-54816978 on Build GRCh38
rs757287929 Chr.1: 54817118-54817118 on Build GRCh38
rs148430708 Chr.1: 54817107-54817107 on Build GRCh38
rs4999211 Chr.1: 54817068-54817068 on Build GRCh38
rs781171383 Chr.1: 54817127-54817127 on Build GRCh38
rs781628316 Chr.1: 54817052-54817052 on Build GRCh38
rs989208592 Chr.1: 54817053-54817053 on Build GRCh38
rs1051071602 Chr.1: 54817017-54817017 on Build GRCh38
rs762699773 Chr.1: 54817154-54817154 on Build GRCh38
rs770540851 Chr.1: 54817160-54817160 on Build GRCh38
rs766014289 Chr.1: 54817082-54817082 on Build GRCh38
rs927900802 Chr.1: 54817085-54817085 on Build GRCh38
rs79893571 Chr.1: 54817155-54817155 on Build GRCh38
rs759421660 Chr.1: 54817163-54817164 on Build GRCh38
rs369275116 Chr.1: 54816976-54816976 on Build GRCh38
rs376402163 Chr.1: 54817106-54817106 on Build GRCh38
rs146890884 Chr.1: 54817046-54817046 on Build GRCh38
rs372600224 Chr.1: 54817142-54817142 on Build GRCh38
rs955488588 Chr.1: 54817164-54817164 on Build GRCh38
rs758992337 Chr.1: 54816974-54816974 on Build GRCh38
rs957787005 Chr.1: 54817146-54817146 on Build GRCh38
rs765021611 Chr.1: 54817169-54817169 on Build GRCh38
rs886409113 Chr.1: 54817109-54817109 on Build GRCh38
rs766161254 Chr.1: 54817000-54817001 on Build GRCh38
rs756645215 Chr.1: 54817138-54817138 on Build GRCh38
rs375175699 Chr.1: 54817029-54817029 on Build GRCh38
rs754760108 Chr.1: 54817008-54817008 on Build GRCh38
rs267598657 Chr.1: 54816988-54816988 on Build GRCh38
rs758681050 Chr.1: 54817045-54817045 on Build GRCh38
rs1029231831 Chr.1: 54817024-54817024 on Build GRCh38
rs747029211 Chr.1: 54817047-54817047 on Build GRCh38
rs144772293 Chr.1: 54817007-54817007 on Build GRCh38
rs986758873 Chr.1: 54817166-54817166 on Build GRCh38
rs764247458 Chr.1: 54817113-54817113 on Build GRCh38
rs751803079 Chr.1: 54817117-54817117 on Build GRCh38
rs373489791 Chr.1: 54817064-54817064 on Build GRCh38
rs769160828 Chr.1: 54817147-54817147 on Build GRCh38
rs191593952 Chr.1: 54817161-54817161 on Build GRCh38
rs143295653 Chr.1: 54817058-54817058 on Build GRCh38
rs899314672 Chr.1: 54817000-54817000 on Build GRCh38
rs562564414 Chr.1: 54816992-54816992 on Build GRCh38
rs747831975 Chr.1: 54817059-54817059 on Build GRCh38
rs752943847 Chr.1: 54817040-54817040 on Build GRCh38

View Full Product Details