Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 34623861-34624086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARID3C
Entrez Gene ID
138715
Gene Name
AT-rich interaction domain 3C
Gene Aliases
-
UniGene
Hs.534549
Gene Chromosome Location
Chr.9: 34621360-34633112 on Build GRCh38
Transcripts
NM_001017363.1 XM_017014283.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2773402 c.407G>A p.R136H Chr.9: 34624032-34624032 on Build GRCh38
6502912 c.395C>T p.T132M Chr.9: 34624044-34624044 on Build GRCh38
6230628 c.421G>A p.A141T Chr.9: 34624018-34624018 on Build GRCh38
4422212 c.429G>T p.Q143H Chr.9: 34624010-34624010 on Build GRCh38
6530605 c.399A>G p.P133P Chr.9: 34624040-34624040 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753149236 Chr.9: 34623965-34623965 on Build GRCh38
rs750449475 Chr.9: 34624010-34624010 on Build GRCh38
rs867863357 Chr.9: 34624000-34624000 on Build GRCh38
rs751753753 Chr.9: 34623883-34623883 on Build GRCh38
rs781355051 Chr.9: 34624009-34624009 on Build GRCh38
rs772552481 Chr.9: 34623953-34623953 on Build GRCh38
rs768367505 Chr.9: 34623931-34623931 on Build GRCh38
rs773982230 Chr.9: 34623932-34623932 on Build GRCh38
rs142104753 Chr.9: 34624022-34624022 on Build GRCh38
rs751843571 Chr.9: 34623991-34623991 on Build GRCh38
rs761401108 Chr.9: 34623935-34623935 on Build GRCh38
rs1005237429 Chr.9: 34623945-34623945 on Build GRCh38
rs372986979 Chr.9: 34623958-34623958 on Build GRCh38
rs769808378 Chr.9: 34624030-34624031 on Build GRCh38
rs745850068 Chr.9: 34623893-34623893 on Build GRCh38
rs1038069619 Chr.9: 34624007-34624007 on Build GRCh38
rs147577275 Chr.9: 34623929-34623929 on Build GRCh38
rs745852051 Chr.9: 34623990-34623991 on Build GRCh38
rs567205717 Chr.9: 34623980-34623980 on Build GRCh38
rs1010395510 Chr.9: 34623927-34623927 on Build GRCh38
rs771619874 Chr.9: 34623947-34623947 on Build GRCh38
rs764466864 Chr.9: 34623984-34623984 on Build GRCh38
rs768457577 Chr.9: 34624034-34624034 on Build GRCh38
rs779187440 Chr.9: 34624059-34624060 on Build GRCh38
rs749142445 Chr.9: 34624033-34624033 on Build GRCh38
rs779950126 Chr.9: 34623917-34623917 on Build GRCh38
rs756119908 Chr.9: 34624016-34624016 on Build GRCh38
rs757346189 Chr.9: 34623886-34623886 on Build GRCh38
rs376209558 Chr.9: 34623889-34623889 on Build GRCh38
rs769569665 Chr.9: 34623894-34623894 on Build GRCh38
rs886780224 Chr.9: 34623919-34623919 on Build GRCh38
rs757382174 Chr.9: 34623995-34623995 on Build GRCh38
rs780790532 Chr.9: 34623931-34623932 on Build GRCh38
rs760106650 Chr.9: 34623956-34623956 on Build GRCh38
rs1036557791 Chr.9: 34623987-34623987 on Build GRCh38

View Full Product Details