Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 9428640-9428901 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPSF3
Entrez Gene ID
51692
Gene Name
cleavage and polyadenylation specific factor 3
Gene Aliases
CPSF-73, CPSF73
UniGene
Hs.515972
Gene Chromosome Location
Chr.2: 9423537-9473110 on Build GRCh38
Transcripts
NM_001321833.1 NM_001321834.1 NM_001321835.1 NM_001321836.1 NM_016207.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7094774 c.88C>A p.L30I Chr.2: 9428802-9428802 on Build GRCh38
392717 c.58delG p.Q21fs*3 Chr.2: 9428772-9428772 on Build GRCh38
1615218 c.114+10A>G p.? Chr.2: 9428838-9428838 on Build GRCh38
4834700 c.101G>A p.G34E Chr.2: 9428815-9428815 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs560603740 Chr.2: 9428698-9428698 on Build GRCh38
rs184756158 Chr.2: 9428683-9428683 on Build GRCh38
rs1048594813 Chr.2: 9428861-9428861 on Build GRCh38
rs199919455 Chr.2: 9428736-9428736 on Build GRCh38
rs762717910 Chr.2: 9428802-9428802 on Build GRCh38
rs149112325 Chr.2: 9428705-9428705 on Build GRCh38
rs779703604 Chr.2: 9428724-9428724 on Build GRCh38
rs145087773 Chr.2: 9428688-9428688 on Build GRCh38
rs768772398 Chr.2: 9428757-9428757 on Build GRCh38
rs371012899 Chr.2: 9428839-9428839 on Build GRCh38
rs906038974 Chr.2: 9428754-9428754 on Build GRCh38
rs757805833 Chr.2: 9428723-9428724 on Build GRCh38
rs761517160 Chr.2: 9428769-9428769 on Build GRCh38
rs750067417 Chr.2: 9428800-9428800 on Build GRCh38
rs774610069 Chr.2: 9428748-9428748 on Build GRCh38
rs767583296 Chr.2: 9428697-9428697 on Build GRCh38
rs1039220210 Chr.2: 9428764-9428764 on Build GRCh38
rs572116806 Chr.2: 9428686-9428686 on Build GRCh38
rs970334105 Chr.2: 9428667-9428667 on Build GRCh38
rs768294046 Chr.2: 9428849-9428849 on Build GRCh38
rs759681062 Chr.2: 9428750-9428750 on Build GRCh38
rs746606768 Chr.2: 9428747-9428747 on Build GRCh38
rs532163240 Chr.2: 9428846-9428846 on Build GRCh38
rs771032861 Chr.2: 9428743-9428743 on Build GRCh38
rs533253680 Chr.2: 9428675-9428675 on Build GRCh38
rs921441469 Chr.2: 9428827-9428827 on Build GRCh38
rs757511050 Chr.2: 9428699-9428699 on Build GRCh38
rs866660801 Chr.2: 9428789-9428789 on Build GRCh38
rs753183078 Chr.2: 9428847-9428847 on Build GRCh38
rs143561900 Chr.2: 9428804-9428804 on Build GRCh38
rs76512430 Chr.2: 9428781-9428781 on Build GRCh38
rs867970471 Chr.2: 9428773-9428773 on Build GRCh38
rs778246755 Chr.2: 9428867-9428867 on Build GRCh38
rs776652022 Chr.2: 9428765-9428765 on Build GRCh38
rs751140543 Chr.2: 9428817-9428817 on Build GRCh38
rs932724587 Chr.2: 9428669-9428669 on Build GRCh38
rs374625647 Chr.2: 9428855-9428855 on Build GRCh38
rs781596368 Chr.2: 9428859-9428859 on Build GRCh38
rs750577595 Chr.2: 9428838-9428838 on Build GRCh38

View Full Product Details