Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49337810-49338065 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD37
Entrez Gene ID
951
Gene Name
CD37 molecule
Gene Aliases
GP52-40, TSPAN26
UniGene
Hs.166556
Gene Chromosome Location
Chr.19: 49335314-49340608 on Build GRCh38
Transcripts
NM_001040031.1 NM_001774.2 XM_005259435.3 XM_005259436.3 XM_011527542.2 XM_011527543.2 XM_011527544.2 XM_017027513.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2752391 c.413C>A p.A138E Chr.19: 49337995-49337995 on Build GRCh38
4941075 c.432C>G p.D144E Chr.19: 49338014-49338014 on Build GRCh38
5343387 c.401C>A p.P134H Chr.19: 49337983-49337983 on Build GRCh38
6623733 c.412G>A p.A138T Chr.19: 49337994-49337994 on Build GRCh38
4983455 c.385A>G p.K129E Chr.19: 49337967-49337967 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142803870 Chr.19: 49337923-49337923 on Build GRCh38
rs753938492 Chr.19: 49338030-49338031 on Build GRCh38
rs374716933 Chr.19: 49337882-49337882 on Build GRCh38
rs757976824 Chr.19: 49338000-49338000 on Build GRCh38
rs763287483 Chr.19: 49337984-49337984 on Build GRCh38
rs760918965 Chr.19: 49337952-49337952 on Build GRCh38
rs201304662 Chr.19: 49338025-49338025 on Build GRCh38
rs374202771 Chr.19: 49338017-49338017 on Build GRCh38
rs371469311 Chr.19: 49338003-49338003 on Build GRCh38
rs202208108 Chr.19: 49337906-49337906 on Build GRCh38
rs562885841 Chr.19: 49337883-49337883 on Build GRCh38
rs753044714 Chr.19: 49337983-49337997 on Build GRCh38
rs780703596 Chr.19: 49338001-49338001 on Build GRCh38
rs750757032 Chr.19: 49337921-49337921 on Build GRCh38
rs779642840 Chr.19: 49337927-49337927 on Build GRCh38
rs377403465 Chr.19: 49337946-49337946 on Build GRCh38
rs372862035 Chr.19: 49337856-49337856 on Build GRCh38
rs956936385 Chr.19: 49337840-49337840 on Build GRCh38
rs888406921 Chr.19: 49337969-49337969 on Build GRCh38
rs780652881 Chr.19: 49337930-49337930 on Build GRCh38
rs757795293 Chr.19: 49337900-49337900 on Build GRCh38
rs769983912 Chr.19: 49338040-49338040 on Build GRCh38
rs749146543 Chr.19: 49337948-49337948 on Build GRCh38
rs754499808 Chr.19: 49337929-49337929 on Build GRCh38
rs752147029 Chr.19: 49338011-49338011 on Build GRCh38
rs370552650 Chr.19: 49337949-49337949 on Build GRCh38
rs754436689 Chr.19: 49337887-49337887 on Build GRCh38
rs769671145 Chr.19: 49337945-49337945 on Build GRCh38
rs914901594 Chr.19: 49337854-49337854 on Build GRCh38
rs764107781 Chr.19: 49337972-49337972 on Build GRCh38
rs989145843 Chr.19: 49337842-49337842 on Build GRCh38
rs183980646 Chr.19: 49337981-49337981 on Build GRCh38
rs1024157704 Chr.19: 49337916-49337916 on Build GRCh38
rs775623827 Chr.19: 49337950-49337950 on Build GRCh38
rs147415665 Chr.19: 49337973-49337973 on Build GRCh38
rs377763244 Chr.19: 49337886-49337886 on Build GRCh38
rs768938080 Chr.19: 49338035-49338035 on Build GRCh38
rs749225161 Chr.19: 49338019-49338019 on Build GRCh38
rs760721886 Chr.19: 49337884-49337884 on Build GRCh38
rs772951476 Chr.19: 49337879-49337879 on Build GRCh38
rs748115500 Chr.19: 49337940-49337940 on Build GRCh38
rs1054356504 Chr.19: 49337855-49337855 on Build GRCh38
rs367859246 Chr.19: 49337928-49337928 on Build GRCh38
rs560803764 Chr.19: 49338012-49338012 on Build GRCh38
rs970797367 Chr.19: 49337943-49337943 on Build GRCh38
rs766747477 Chr.19: 49337995-49337995 on Build GRCh38
rs778729121 Chr.19: 49338026-49338026 on Build GRCh38
rs747214284 Chr.19: 49338030-49338030 on Build GRCh38
rs762049076 Chr.19: 49338039-49338039 on Build GRCh38
rs1050562215 Chr.19: 49337871-49337871 on Build GRCh38
rs375688108 Chr.19: 49337922-49337922 on Build GRCh38
rs199925267 Chr.19: 49338037-49338037 on Build GRCh38

View Full Product Details