Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 22600784-22601031 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8orf58
Entrez Gene ID
541565
Gene Name
chromosome 8 open reading frame 58
Gene Aliases
-
UniGene
Hs.743508
Gene Chromosome Location
Chr.8: 22599601-22604150 on Build GRCh38
Transcripts
NM_001013842.2 NM_001198827.1 NM_173686.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4881470 c.95C>G p.T32S Chr.8: 22600936-22600936 on Build GRCh38
1456109 c.115G>A p.A39T Chr.8: 22600956-22600956 on Build GRCh38
2786282 c.165C>T p.V55V Chr.8: 22601006-22601006 on Build GRCh38
4473725 c.186C>T p.L62L Chr.8: 22601027-22601027 on Build GRCh38
2786281 c.103C>T p.R35W Chr.8: 22600944-22600944 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs997938134 Chr.8: 22600815-22600815 on Build GRCh38
rs770654323 Chr.8: 22600843-22600843 on Build GRCh38
rs767622420 Chr.8: 22600865-22600865 on Build GRCh38
rs953795920 Chr.8: 22600821-22600821 on Build GRCh38
rs138371466 Chr.8: 22601006-22601006 on Build GRCh38
rs978364857 Chr.8: 22600862-22600862 on Build GRCh38
rs750822224 Chr.8: 22601003-22601003 on Build GRCh38
rs1021947281 Chr.8: 22600842-22600842 on Build GRCh38
rs761235675 Chr.8: 22600893-22600893 on Build GRCh38
rs145988500 Chr.8: 22600944-22600944 on Build GRCh38
rs749215293 Chr.8: 22600950-22600950 on Build GRCh38
rs746100904 Chr.8: 22600956-22600956 on Build GRCh38
rs916770272 Chr.8: 22600925-22600925 on Build GRCh38
rs200708476 Chr.8: 22600952-22600952 on Build GRCh38
rs372629327 Chr.8: 22600837-22600837 on Build GRCh38
rs897036841 Chr.8: 22600965-22600965 on Build GRCh38
rs748636813 Chr.8: 22600819-22600819 on Build GRCh38
rs370995356 Chr.8: 22601000-22601000 on Build GRCh38
rs992289672 Chr.8: 22600918-22600918 on Build GRCh38
rs924223904 Chr.8: 22600874-22600874 on Build GRCh38
rs557361550 Chr.8: 22600964-22600964 on Build GRCh38
rs150454220 Chr.8: 22600945-22600945 on Build GRCh38
rs909199007 Chr.8: 22600946-22600946 on Build GRCh38
rs376856636 Chr.8: 22600855-22600855 on Build GRCh38
rs1036326035 Chr.8: 22600957-22600957 on Build GRCh38
rs948135032 Chr.8: 22600935-22600935 on Build GRCh38
rs754422545 Chr.8: 22600905-22600905 on Build GRCh38
rs764305189 Chr.8: 22600972-22600972 on Build GRCh38
rs181513488 Chr.8: 22600929-22600929 on Build GRCh38
rs764750697 Chr.8: 22600902-22600902 on Build GRCh38
rs755987455 Chr.8: 22600937-22600937 on Build GRCh38
rs751163163 Chr.8: 22600923-22600923 on Build GRCh38
rs868537785 Chr.8: 22600976-22600976 on Build GRCh38
rs759598085 Chr.8: 22600888-22600888 on Build GRCh38
rs201978138 Chr.8: 22601001-22601001 on Build GRCh38
rs754642651 Chr.8: 22600930-22600930 on Build GRCh38
rs913282878 Chr.8: 22600839-22600839 on Build GRCh38
rs578114033 Chr.8: 22600872-22600872 on Build GRCh38
rs765617757 Chr.8: 22601002-22601002 on Build GRCh38
rs767208210 Chr.8: 22601007-22601007 on Build GRCh38
rs772226148 Chr.8: 22600961-22600961 on Build GRCh38
rs757606071 Chr.8: 22600906-22600906 on Build GRCh38
rs760791712 Chr.8: 22600969-22600969 on Build GRCh38
rs998075395 Chr.8: 22601004-22601004 on Build GRCh38
rs759692286 Chr.8: 22600856-22600856 on Build GRCh38
rs774073103 Chr.8: 22600955-22600955 on Build GRCh38
rs780673336 Chr.8: 22600932-22600932 on Build GRCh38
rs747695605 Chr.8: 22600940-22600940 on Build GRCh38
rs887448224 Chr.8: 22600978-22600978 on Build GRCh38
rs896982917 Chr.8: 22600809-22600809 on Build GRCh38
rs960957230 Chr.8: 22600894-22600894 on Build GRCh38

View Full Product Details