Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218814179-218814418 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP27A1
Entrez Gene ID
1593
Gene Name
cytochrome P450 family 27 subfamily A member 1
Gene Aliases
CP27, CTX, CYP27
UniGene
Hs.516700
Gene Chromosome Location
Chr.2: 218781006-218815293 on Build GRCh38
Transcripts
NM_000784.3 XM_017003488.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7363514 c.1183C>A p.R395S Chr.2: 218814186-218814186 on Build GRCh38
4933266 c.1213C>T p.R405W Chr.2: 218814408-218814408 on Build GRCh38
1326413 c.1220T>C p.I407T Chr.2: 218814415-218814415 on Build GRCh38
6010649 c.1185-6C>T p.? Chr.2: 218814374-218814374 on Build GRCh38
4954681 c.1218C>G p.I406M Chr.2: 218814413-218814413 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs117727806 Chr.2: 218814365-218814365 on Build GRCh38
rs587778779 Chr.2: 218814379-218814379 on Build GRCh38
rs751088394 Chr.2: 218814201-218814201 on Build GRCh38
rs34494019 Chr.2: 218814316-218814317 on Build GRCh38
rs772157208 Chr.2: 218814367-218814367 on Build GRCh38
rs756771182 Chr.2: 218814212-218814212 on Build GRCh38
rs587778780 Chr.2: 218814397-218814397 on Build GRCh38
rs1042072855 Chr.2: 218814213-218814213 on Build GRCh38
rs780338051 Chr.2: 218814337-218814337 on Build GRCh38
rs1007682792 Chr.2: 218814259-218814259 on Build GRCh38
rs368189284 Chr.2: 218814335-218814335 on Build GRCh38
rs997846128 Chr.2: 218814232-218814232 on Build GRCh38
rs73991002 Chr.2: 218814242-218814242 on Build GRCh38
rs779178036 Chr.2: 218814364-218814364 on Build GRCh38
rs781590975 Chr.2: 218814229-218814229 on Build GRCh38
rs540682832 Chr.2: 218814361-218814361 on Build GRCh38
rs373628198 Chr.2: 218814377-218814377 on Build GRCh38
rs768823291 Chr.2: 218814345-218814345 on Build GRCh38
rs756491129 Chr.2: 218814336-218814336 on Build GRCh38
rs774575679 Chr.2: 218814245-218814245 on Build GRCh38
rs889528476 Chr.2: 218814251-218814251 on Build GRCh38
rs776459243 Chr.2: 218814376-218814376 on Build GRCh38
rs1803361 Chr.2: 218814394-218814394 on Build GRCh38
rs761941429 Chr.2: 218814334-218814335 on Build GRCh38
rs575111058 Chr.2: 218814209-218814209 on Build GRCh38
rs949118725 Chr.2: 218814328-218814328 on Build GRCh38
rs902199827 Chr.2: 218814228-218814228 on Build GRCh38
rs917554209 Chr.2: 218814296-218814296 on Build GRCh38

View Full Product Details