Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 149572148-149572401 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GINM1
Entrez Gene ID
116254
Gene Name
glycoprotein integral membrane 1
Gene Aliases
C6orf72, dJ12G14.2
UniGene
Hs.438872
Gene Chromosome Location
Chr.6: 149566392-149590931 on Build GRCh38
Transcripts
NM_138785.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
421040 c.169T>C p.S57P Chr.6: 149572333-149572333 on Build GRCh38
6671033 c.129C>T p.I43I Chr.6: 149572293-149572293 on Build GRCh38
1074430 c.124G>A p.G42S Chr.6: 149572288-149572288 on Build GRCh38
740656 c.147A>C p.T49T Chr.6: 149572311-149572311 on Build GRCh38
1074429 c.122A>C p.D41A Chr.6: 149572286-149572286 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs376235739 Chr.6: 149572287-149572287 on Build GRCh38
rs923295469 Chr.6: 149572195-149572195 on Build GRCh38
rs771917817 Chr.6: 149572322-149572322 on Build GRCh38
rs766516085 Chr.6: 149572248-149572248 on Build GRCh38
rs771172096 Chr.6: 149572333-149572333 on Build GRCh38
rs1042273577 Chr.6: 149572311-149572311 on Build GRCh38
rs370032821 Chr.6: 149572282-149572282 on Build GRCh38
rs147898628 Chr.6: 149572228-149572228 on Build GRCh38
rs144747991 Chr.6: 149572194-149572194 on Build GRCh38
rs199901540 Chr.6: 149572307-149572307 on Build GRCh38
rs753242749 Chr.6: 149572265-149572265 on Build GRCh38
rs747124651 Chr.6: 149572330-149572330 on Build GRCh38
rs374063431 Chr.6: 149572370-149572370 on Build GRCh38
rs568062561 Chr.6: 149572227-149572227 on Build GRCh38
rs754864124 Chr.6: 149572289-149572289 on Build GRCh38
rs776976102 Chr.6: 149572250-149572250 on Build GRCh38
rs954907550 Chr.6: 149572198-149572198 on Build GRCh38
rs751588704 Chr.6: 149572241-149572242 on Build GRCh38
rs758858567 Chr.6: 149572269-149572269 on Build GRCh38
rs947324550 Chr.6: 149572231-149572231 on Build GRCh38
rs376442666 Chr.6: 149572240-149572240 on Build GRCh38
rs113794184 Chr.6: 149572255-149572255 on Build GRCh38
rs937876714 Chr.6: 149572238-149572238 on Build GRCh38
rs748052857 Chr.6: 149572310-149572310 on Build GRCh38
rs776886244 Chr.6: 149572342-149572342 on Build GRCh38
rs1026404932 Chr.6: 149572193-149572193 on Build GRCh38
rs554691536 Chr.6: 149572196-149572196 on Build GRCh38
rs142853295 Chr.6: 149572288-149572288 on Build GRCh38
rs377477084 Chr.6: 149572271-149572271 on Build GRCh38
rs372681349 Chr.6: 149572260-149572260 on Build GRCh38
rs773278724 Chr.6: 149572326-149572326 on Build GRCh38

View Full Product Details