Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74544522-74544781 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD300C
Entrez Gene ID
10871
Gene Name
CD300c molecule
Gene Aliases
CLM-6, CMRF-35, CMRF-35A, CMRF35, CMRF35-A1, CMRF35A, CMRF35A1, IGSF16, LIR
UniGene
Hs.2605
Gene Chromosome Location
Chr.17: 74534359-74546171 on Build GRCh38
Transcripts
NM_006678.4 XM_017024033.1 XM_017024034.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
300586 c.383A>T p.E128V Chr.17: 74544626-74544626 on Build GRCh38
6404444 c.353G>A p.R118Q Chr.17: 74544656-74544656 on Build GRCh38
6408525 c.370A>G p.I124V Chr.17: 74544639-74544639 on Build GRCh38
4842915 c.382G>C p.E128Q Chr.17: 74544627-74544627 on Build GRCh38
6623676 c.352C>T p.R118* Chr.17: 74544657-74544657 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146418916 Chr.17: 74544759-74544759 on Build GRCh38
rs965041395 Chr.17: 74544606-74544606 on Build GRCh38
rs773791098 Chr.17: 74544565-74544565 on Build GRCh38
rs374234481 Chr.17: 74544665-74544665 on Build GRCh38
rs759818914 Chr.17: 74544664-74544664 on Build GRCh38
rs370897171 Chr.17: 74544629-74544629 on Build GRCh38
rs760499467 Chr.17: 74544610-74544610 on Build GRCh38
rs200819404 Chr.17: 74544684-74544684 on Build GRCh38
rs530341053 Chr.17: 74544697-74544697 on Build GRCh38
rs778044136 Chr.17: 74544633-74544633 on Build GRCh38
rs746663607 Chr.17: 74544730-74544730 on Build GRCh38
rs201636315 Chr.17: 74544690-74544690 on Build GRCh38
rs769605668 Chr.17: 74544756-74544756 on Build GRCh38
rs546382422 Chr.17: 74544619-74544619 on Build GRCh38
rs866975838 Chr.17: 74544702-74544702 on Build GRCh38
rs779759691 Chr.17: 74544727-74544727 on Build GRCh38
rs879594489 Chr.17: 74544554-74544554 on Build GRCh38
rs144695162 Chr.17: 74544657-74544657 on Build GRCh38
rs745316727 Chr.17: 74544647-74544647 on Build GRCh38
rs374552644 Chr.17: 74544680-74544680 on Build GRCh38
rs763827378 Chr.17: 74544611-74544611 on Build GRCh38
rs754082658 Chr.17: 74544693-74544693 on Build GRCh38
rs775297353 Chr.17: 74544597-74544597 on Build GRCh38
rs759568586 Chr.17: 74544673-74544673 on Build GRCh38
rs111380989 Chr.17: 74544656-74544656 on Build GRCh38
rs779564893 Chr.17: 74544642-74544642 on Build GRCh38
rs776352764 Chr.17: 74544744-74544744 on Build GRCh38
rs747843328 Chr.17: 74544745-74544745 on Build GRCh38
rs944815734 Chr.17: 74544613-74544613 on Build GRCh38
rs966418855 Chr.17: 74544721-74544721 on Build GRCh38
rs1017705752 Chr.17: 74544580-74544580 on Build GRCh38
rs771783559 Chr.17: 74544592-74544592 on Build GRCh38
rs990669343 Chr.17: 74544550-74544550 on Build GRCh38
rs771730864 Chr.17: 74544648-74544648 on Build GRCh38
rs756347680 Chr.17: 74544689-74544689 on Build GRCh38
rs759256102 Chr.17: 74544577-74544577 on Build GRCh38
rs767674440 Chr.17: 74544624-74544624 on Build GRCh38
rs376507412 Chr.17: 74544608-74544608 on Build GRCh38
rs368005305 Chr.17: 74544732-74544732 on Build GRCh38
rs749787461 Chr.17: 74544634-74544634 on Build GRCh38
rs374478609 Chr.17: 74544709-74544709 on Build GRCh38
rs761896670 Chr.17: 74544727-74544728 on Build GRCh38
rs770452471 Chr.17: 74544561-74544561 on Build GRCh38
rs199715562 Chr.17: 74544700-74544700 on Build GRCh38
rs776351124 Chr.17: 74544660-74544660 on Build GRCh38
rs752946104 Chr.17: 74544625-74544625 on Build GRCh38
rs140589759 Chr.17: 74544652-74544652 on Build GRCh38
rs547924214 Chr.17: 74544623-74544623 on Build GRCh38
rs953974833 Chr.17: 74544738-74544738 on Build GRCh38
rs757771063 Chr.17: 74544639-74544639 on Build GRCh38
rs1032111265 Chr.17: 74544734-74544734 on Build GRCh38
rs565275112 Chr.17: 74544618-74544618 on Build GRCh38
rs773149738 Chr.17: 74544667-74544667 on Build GRCh38
rs1049243246 Chr.17: 74544638-74544638 on Build GRCh38
rs1020693915 Chr.17: 74544696-74544696 on Build GRCh38

View Full Product Details