Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 56945924-56946153 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8B
Entrez Gene ID
732
Gene Name
complement component 8, beta polypeptide
Gene Aliases
C82
UniGene
Hs.391835
Gene Chromosome Location
Chr.1: 56929210-56974383 on Build GRCh38
Transcripts
NM_000066.3 NM_001278543.1 NM_001278544.1 XM_017002235.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4972332 c.880C>A p.H294N Chr.1: 56946046-56946046 on Build GRCh38
6875991 c.978G>A p.L326L Chr.1: 56945948-56945948 on Build GRCh38
911104 c.990C>T p.Y330Y Chr.1: 56945936-56945936 on Build GRCh38
6126818 c.945C>A p.Y315* Chr.1: 56945981-56945981 on Build GRCh38
911105 c.884C>T p.A295V Chr.1: 56946042-56946042 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs545706495 Chr.1: 56946018-56946018 on Build GRCh38
rs776510959 Chr.1: 56946072-56946072 on Build GRCh38
rs761418803 Chr.1: 56945981-56945981 on Build GRCh38
rs149538771 Chr.1: 56946021-56946021 on Build GRCh38
rs548087620 Chr.1: 56946055-56946055 on Build GRCh38
rs753647247 Chr.1: 56946005-56946005 on Build GRCh38
rs147180727 Chr.1: 56946040-56946040 on Build GRCh38
rs771548491 Chr.1: 56945965-56945965 on Build GRCh38
rs763148835 Chr.1: 56946081-56946081 on Build GRCh38
rs994766444 Chr.1: 56946113-56946113 on Build GRCh38
rs779329101 Chr.1: 56946093-56946093 on Build GRCh38
rs141429572 Chr.1: 56945996-56945996 on Build GRCh38
rs746172125 Chr.1: 56945971-56945971 on Build GRCh38
rs753752153 Chr.1: 56946094-56946094 on Build GRCh38
rs369071394 Chr.1: 56946071-56946071 on Build GRCh38
rs751704682 Chr.1: 56946102-56946102 on Build GRCh38
rs41286852 Chr.1: 56946106-56946106 on Build GRCh38
rs755983290 Chr.1: 56946020-56946020 on Build GRCh38
rs781743688 Chr.1: 56946039-56946039 on Build GRCh38
rs372123388 Chr.1: 56946057-56946057 on Build GRCh38
rs764986723 Chr.1: 56946074-56946074 on Build GRCh38
rs200236866 Chr.1: 56946080-56946080 on Build GRCh38
rs375053307 Chr.1: 56946066-56946066 on Build GRCh38
rs748280435 Chr.1: 56945953-56945953 on Build GRCh38
rs369652003 Chr.1: 56945960-56945960 on Build GRCh38
rs754717773 Chr.1: 56946011-56946011 on Build GRCh38
rs867625308 Chr.1: 56946103-56946103 on Build GRCh38
rs374138669 Chr.1: 56946031-56946031 on Build GRCh38
rs141295453 Chr.1: 56945959-56945959 on Build GRCh38
rs771744350 Chr.1: 56946056-56946056 on Build GRCh38
rs201463530 Chr.1: 56945992-56945992 on Build GRCh38
rs758366170 Chr.1: 56945958-56945958 on Build GRCh38
rs146176997 Chr.1: 56945969-56945969 on Build GRCh38
rs1027535561 Chr.1: 56946114-56946114 on Build GRCh38
rs760664100 Chr.1: 56945997-56945997 on Build GRCh38
rs945021500 Chr.1: 56946061-56946061 on Build GRCh38
rs200297171 Chr.1: 56946073-56946073 on Build GRCh38
rs968552522 Chr.1: 56945986-56945986 on Build GRCh38
rs775893210 Chr.1: 56946053-56946053 on Build GRCh38
rs770244658 Chr.1: 56946050-56946050 on Build GRCh38
rs201686818 Chr.1: 56945980-56945980 on Build GRCh38
rs781075590 Chr.1: 56946048-56946048 on Build GRCh38
rs534197740 Chr.1: 56946029-56946029 on Build GRCh38
rs944563967 Chr.1: 56946091-56946091 on Build GRCh38
rs371944655 Chr.1: 56945998-56945998 on Build GRCh38
rs919007642 Chr.1: 56946117-56946117 on Build GRCh38

View Full Product Details