Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 120739183-120739414 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACADS
Entrez Gene ID
35
Gene Name
acyl-CoA dehydrogenase, C-2 to C-3 short chain
Gene Aliases
ACAD3, SCAD
UniGene
Hs.507076
Gene Chromosome Location
Chr.12: 120725738-120740008 on Build GRCh38
Transcripts
NM_000017.3 NM_001302554.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3457074 c.1129C>G p.P377A Chr.12: 120739338-120739338 on Build GRCh38
5913318 c.1105G>A p.G369S Chr.12: 120739314-120739314 on Build GRCh38
39701 c.1200G>A p.L400L Chr.12: 120739409-120739409 on Build GRCh38
5913317 c.1106G>A p.G369D Chr.12: 120739315-120739315 on Build GRCh38
7222027 c.1094A>G p.Q365R Chr.12: 120739303-120739303 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs895841347 Chr.12: 120739385-120739385 on Build GRCh38
rs368469075 Chr.12: 120739304-120739304 on Build GRCh38
rs775999272 Chr.12: 120739391-120739391 on Build GRCh38
rs773946131 Chr.12: 120739268-120739268 on Build GRCh38
rs776623770 Chr.12: 120739201-120739201 on Build GRCh38
rs766183395 Chr.12: 120739366-120739366 on Build GRCh38
rs201218974 Chr.12: 120739279-120739279 on Build GRCh38
rs370077075 Chr.12: 120739221-120739221 on Build GRCh38
rs953436518 Chr.12: 120739344-120739344 on Build GRCh38
rs762443848 Chr.12: 120739290-120739290 on Build GRCh38
rs771814293 Chr.12: 120739361-120739361 on Build GRCh38
rs145466253 Chr.12: 120739314-120739314 on Build GRCh38
rs1031962576 Chr.12: 120739275-120739275 on Build GRCh38
rs995064724 Chr.12: 120739338-120739338 on Build GRCh38
rs35233375 Chr.12: 120739357-120739357 on Build GRCh38
rs202124189 Chr.12: 120739362-120739362 on Build GRCh38
rs566325901 Chr.12: 120739317-120739317 on Build GRCh38
rs775074247 Chr.12: 120739287-120739287 on Build GRCh38
rs371934737 Chr.12: 120739326-120739326 on Build GRCh38
rs368572195 Chr.12: 120739227-120739227 on Build GRCh38
rs751801455 Chr.12: 120739217-120739217 on Build GRCh38
rs1057516421 Chr.12: 120739373-120739374 on Build GRCh38
rs371323002 Chr.12: 120739358-120739358 on Build GRCh38
rs183161718 Chr.12: 120739339-120739339 on Build GRCh38
rs948685149 Chr.12: 120739392-120739392 on Build GRCh38
rs28940872 Chr.12: 120739356-120739356 on Build GRCh38
rs751738987 Chr.12: 120739315-120739315 on Build GRCh38
rs769675520 Chr.12: 120739254-120739254 on Build GRCh38
rs750909218 Chr.12: 120739310-120739310 on Build GRCh38
rs28940875 Chr.12: 120739347-120739347 on Build GRCh38
rs374867417 Chr.12: 120739340-120739340 on Build GRCh38
rs936141551 Chr.12: 120739244-120739244 on Build GRCh38
rs759774887 Chr.12: 120739394-120739394 on Build GRCh38
rs374392992 Chr.12: 120739225-120739225 on Build GRCh38
rs752240673 Chr.12: 120739221-120739221 on Build GRCh38
rs763455552 Chr.12: 120739381-120739381 on Build GRCh38
rs766657084 Chr.12: 120739382-120739382 on Build GRCh38
rs759217705 Chr.12: 120739274-120739274 on Build GRCh38
rs773646063 Chr.12: 120739378-120739378 on Build GRCh38
rs1015232355 Chr.12: 120739325-120739325 on Build GRCh38
rs537072819 Chr.12: 120739365-120739365 on Build GRCh38
rs200165866 Chr.12: 120739313-120739313 on Build GRCh38
rs371604145 Chr.12: 120739230-120739230 on Build GRCh38
rs953695669 Chr.12: 120739307-120739307 on Build GRCh38
rs926163707 Chr.12: 120739213-120739213 on Build GRCh38
rs561806153 Chr.12: 120739207-120739207 on Build GRCh38
rs368064268 Chr.12: 120739348-120739348 on Build GRCh38
rs796051905 Chr.12: 120739321-120739321 on Build GRCh38
rs770730868 Chr.12: 120739261-120739261 on Build GRCh38
rs777643097 Chr.12: 120739257-120739257 on Build GRCh38
rs780178412 Chr.12: 120739211-120739211 on Build GRCh38
rs758765936 Chr.12: 120739208-120739208 on Build GRCh38
rs550519759 Chr.12: 120739258-120739258 on Build GRCh38

View Full Product Details