Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 102685767-102686040 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL3
Entrez Gene ID
403
Gene Name
ADP ribosylation factor like GTPase 3
Gene Aliases
ARFL3
UniGene
Hs.182215
Gene Chromosome Location
Chr.10: 102673727-102714433 on Build GRCh38
Transcripts
NM_004311.3 XM_017016260.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7232516 c.448G>C p.D150H Chr.10: 102685869-102685869 on Build GRCh38
4011306 c.477A>G p.S159S Chr.10: 102685840-102685840 on Build GRCh38
683520 c.408C>T p.A136A Chr.10: 102685909-102685909 on Build GRCh38
4639296 c.448G>A p.D150N Chr.10: 102685869-102685869 on Build GRCh38
3806359 c.412G>C p.E138Q Chr.10: 102685905-102685905 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757709273 Chr.10: 102685788-102685788 on Build GRCh38
rs538533205 Chr.10: 102685999-102685999 on Build GRCh38
rs763060317 Chr.10: 102686004-102686004 on Build GRCh38
rs752095202 Chr.10: 102685950-102685950 on Build GRCh38
rs754624331 Chr.10: 102685925-102685925 on Build GRCh38
rs767828413 Chr.10: 102685914-102685914 on Build GRCh38
rs201771386 Chr.10: 102685913-102685913 on Build GRCh38
rs769888354 Chr.10: 102685882-102685882 on Build GRCh38
rs745708839 Chr.10: 102685995-102685995 on Build GRCh38
rs551366324 Chr.10: 102685964-102685964 on Build GRCh38
rs769860527 Chr.10: 102685998-102685998 on Build GRCh38
rs939117445 Chr.10: 102685811-102685811 on Build GRCh38
rs778655671 Chr.10: 102685939-102685939 on Build GRCh38
rs191578111 Chr.10: 102685789-102685789 on Build GRCh38
rs182194850 Chr.10: 102685824-102685824 on Build GRCh38
rs778332463 Chr.10: 102685845-102685845 on Build GRCh38
rs397718095 Chr.10: 102685790-102685790 on Build GRCh38
rs896886256 Chr.10: 102685794-102685794 on Build GRCh38
rs150077149 Chr.10: 102685922-102685922 on Build GRCh38
rs374365288 Chr.10: 102685821-102685821 on Build GRCh38
rs763148372 Chr.10: 102685902-102685902 on Build GRCh38
rs753503964 Chr.10: 102685921-102685921 on Build GRCh38
rs770692966 Chr.10: 102685990-102685990 on Build GRCh38
rs746743719 Chr.10: 102685870-102685870 on Build GRCh38
rs376018992 Chr.10: 102685865-102685865 on Build GRCh38
rs971998360 Chr.10: 102685806-102685806 on Build GRCh38
rs370338506 Chr.10: 102685795-102685795 on Build GRCh38
rs776696557 Chr.10: 102685945-102685945 on Build GRCh38
rs757913078 Chr.10: 102685951-102685951 on Build GRCh38
rs5787474 Chr.10: 102685793-102685793 on Build GRCh38
rs571007258 Chr.10: 102685979-102685979 on Build GRCh38
rs770491993 Chr.10: 102685993-102685993 on Build GRCh38
rs547067319 Chr.10: 102685822-102685822 on Build GRCh38
rs759728586 Chr.10: 102685873-102685873 on Build GRCh38
rs3832710 Chr.10: 102685789-102685789 on Build GRCh38
rs777705334 Chr.10: 102685869-102685869 on Build GRCh38
rs894980013 Chr.10: 102685839-102685839 on Build GRCh38
rs748084188 Chr.10: 102685864-102685864 on Build GRCh38
rs764638372 Chr.10: 102685912-102685912 on Build GRCh38
rs368868984 Chr.10: 102685898-102685898 on Build GRCh38
rs755402499 Chr.10: 102686002-102686003 on Build GRCh38
rs770782663 Chr.10: 102685871-102685871 on Build GRCh38
rs776901858 Chr.10: 102685872-102685872 on Build GRCh38

View Full Product Details