Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 120516434-120516681 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COQ5
Entrez Gene ID
84274
Gene Name
coenzyme Q5, methyltransferase
Gene Aliases
-
UniGene
Hs.17250
Gene Chromosome Location
Chr.12: 120503279-120529171 on Build GRCh38
Transcripts
NM_032314.3 XM_006719639.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4795610 c.487G>A p.G163S Chr.12: 120516654-120516654 on Build GRCh38
2175237 c.535G>A p.V179I Chr.12: 120516606-120516606 on Build GRCh38
935919 c.497G>A p.R166H Chr.12: 120516644-120516644 on Build GRCh38
1746799 c.546G>A p.Q182Q Chr.12: 120516595-120516595 on Build GRCh38
467897 c.475G>A p.E159K Chr.12: 120516666-120516666 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139643937 Chr.12: 120516515-120516515 on Build GRCh38
rs747738620 Chr.12: 120516543-120516544 on Build GRCh38
rs761369487 Chr.12: 120516541-120516542 on Build GRCh38
rs143479715 Chr.12: 120516652-120516652 on Build GRCh38
rs763178641 Chr.12: 120516653-120516653 on Build GRCh38
rs778498312 Chr.12: 120516635-120516635 on Build GRCh38
rs561009074 Chr.12: 120516522-120516522 on Build GRCh38
rs769489119 Chr.12: 120516634-120516636 on Build GRCh38
rs752650506 Chr.12: 120516575-120516575 on Build GRCh38
rs766664549 Chr.12: 120516591-120516591 on Build GRCh38
rs866127066 Chr.12: 120516649-120516649 on Build GRCh38
rs745427038 Chr.12: 120516639-120516639 on Build GRCh38
rs376870708 Chr.12: 120516535-120516535 on Build GRCh38
rs751538217 Chr.12: 120516529-120516529 on Build GRCh38
rs776204825 Chr.12: 120516509-120516509 on Build GRCh38
rs184473794 Chr.12: 120516501-120516501 on Build GRCh38
rs770639277 Chr.12: 120516634-120516634 on Build GRCh38
rs748981096 Chr.12: 120516621-120516621 on Build GRCh38
rs201120226 Chr.12: 120516640-120516640 on Build GRCh38
rs755076079 Chr.12: 120516564-120516564 on Build GRCh38
rs756022650 Chr.12: 120516587-120516587 on Build GRCh38
rs192345740 Chr.12: 120516468-120516468 on Build GRCh38
rs529965711 Chr.12: 120516644-120516644 on Build GRCh38
rs997124292 Chr.12: 120516648-120516648 on Build GRCh38
rs543355990 Chr.12: 120516645-120516645 on Build GRCh38
rs1045851364 Chr.12: 120516483-120516483 on Build GRCh38
rs768304940 Chr.12: 120516580-120516580 on Build GRCh38
rs563103401 Chr.12: 120516651-120516651 on Build GRCh38
rs781062422 Chr.12: 120516574-120516574 on Build GRCh38

View Full Product Details