Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 139965316-139965583 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXorf66
Entrez Gene ID
347487
Gene Name
chromosome X open reading frame 66
Gene Aliases
SGPX
UniGene
Hs.121384
Gene Chromosome Location
Chr.X: 139955725-139965518 on Build GRCh38
Transcripts
NM_001013403.2

SNP Detail

SNP ID Chromosome Location
rs369463319 Chr.X: 139965496-139965496 on Build GRCh38
rs748480779 Chr.X: 139965447-139965447 on Build GRCh38
rs372750238 Chr.X: 139965526-139965526 on Build GRCh38
rs762687853 Chr.X: 139965363-139965363 on Build GRCh38
rs199505612 Chr.X: 139965485-139965485 on Build GRCh38
rs778255161 Chr.X: 139965466-139965466 on Build GRCh38
rs1040768401 Chr.X: 139965468-139965468 on Build GRCh38
rs761951922 Chr.X: 139965465-139965465 on Build GRCh38
rs758705464 Chr.X: 139965460-139965460 on Build GRCh38
rs762750661 Chr.X: 139965541-139965541 on Build GRCh38
rs753968776 Chr.X: 139965413-139965413 on Build GRCh38
rs905014749 Chr.X: 139965500-139965500 on Build GRCh38
rs371718733 Chr.X: 139965372-139965372 on Build GRCh38
rs776254666 Chr.X: 139965508-139965508 on Build GRCh38
rs779039398 Chr.X: 139965441-139965441 on Build GRCh38
rs376461807 Chr.X: 139965412-139965412 on Build GRCh38
rs771601810 Chr.X: 139965504-139965504 on Build GRCh38
rs933685226 Chr.X: 139965384-139965384 on Build GRCh38
rs752487723 Chr.X: 139965362-139965362 on Build GRCh38
rs745393036 Chr.X: 139965516-139965516 on Build GRCh38
rs775008122 Chr.X: 139965529-139965529 on Build GRCh38
rs755182392 Chr.X: 139965436-139965436 on Build GRCh38
rs924428134 Chr.X: 139965354-139965354 on Build GRCh38
rs758971586 Chr.X: 139965379-139965379 on Build GRCh38
rs935929560 Chr.X: 139965356-139965356 on Build GRCh38
rs751510341 Chr.X: 139965378-139965378 on Build GRCh38

View Full Product Details