Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150816242-150816469 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARNT
Entrez Gene ID
405
Gene Name
aryl hydrocarbon receptor nuclear translocator
Gene Aliases
HIF-1-beta, HIF-1beta, HIF1-beta, HIF1B, HIF1BETA, TANGO, bHLHe2
UniGene
Hs.632446
Gene Chromosome Location
Chr.1: 150809705-150876768 on Build GRCh38
Transcripts
NM_001197325.1 NM_001286035.1 NM_001286036.1 NM_001668.3 NM_178427.2 XM_005245151.1 XM_005245153.1 XM_005245154.3 XM_005245157.1 XM_011509542.2 XM_011509543.2 XM_011509544.2 XM_011509545.2 XM_011509546.1 XM_011509547.2 XM_017001288.1 XM_017001289.1 XM_017001290.1 XM_017001291.1 XM_017001292.1 XM_017001293.1 XM_017001294.1 XM_017001295.1 XM_017001296.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4532634 c.1900G>A p.G634R Chr.1: 150816309-150816309 on Build GRCh38
1738568 c.1834G>A p.V612I Chr.1: 150816375-150816375 on Build GRCh38
4910168 c.1845A>T p.S615S Chr.1: 150816364-150816364 on Build GRCh38
1747725 c.1836C>G p.V612V Chr.1: 150816373-150816373 on Build GRCh38
1733351 c.1844C>G p.S615* Chr.1: 150816365-150816365 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775019737 Chr.1: 150816369-150816369 on Build GRCh38
rs998245936 Chr.1: 150816349-150816349 on Build GRCh38
rs587718559 Chr.1: 150816420-150816420 on Build GRCh38
rs140420727 Chr.1: 150816272-150816272 on Build GRCh38
rs755503741 Chr.1: 150816306-150816306 on Build GRCh38
rs758403177 Chr.1: 150816289-150816289 on Build GRCh38
rs765561969 Chr.1: 150816404-150816404 on Build GRCh38
rs781761456 Chr.1: 150816438-150816438 on Build GRCh38
rs587624534 Chr.1: 150816279-150816279 on Build GRCh38
rs776420303 Chr.1: 150816395-150816395 on Build GRCh38
rs868189404 Chr.1: 150816332-150816332 on Build GRCh38
rs374089243 Chr.1: 150816320-150816320 on Build GRCh38
rs771464000 Chr.1: 150816358-150816358 on Build GRCh38
rs755557009 Chr.1: 150816437-150816437 on Build GRCh38
rs587640516 Chr.1: 150816405-150816405 on Build GRCh38
rs376139724 Chr.1: 150816331-150816331 on Build GRCh38
rs752088370 Chr.1: 150816297-150816297 on Build GRCh38
rs781694598 Chr.1: 150816314-150816314 on Build GRCh38
rs768511265 Chr.1: 150816380-150816380 on Build GRCh38
rs765314219 Chr.1: 150816282-150816282 on Build GRCh38
rs1006646907 Chr.1: 150816366-150816366 on Build GRCh38
rs778430234 Chr.1: 150816330-150816330 on Build GRCh38
rs760635188 Chr.1: 150816378-150816378 on Build GRCh38
rs767542779 Chr.1: 150816416-150816416 on Build GRCh38
rs889509878 Chr.1: 150816365-150816365 on Build GRCh38
rs754122594 Chr.1: 150816401-150816401 on Build GRCh38
rs867358594 Chr.1: 150816333-150816333 on Build GRCh38
rs753582088 Chr.1: 150816434-150816434 on Build GRCh38
rs750574542 Chr.1: 150816285-150816285 on Build GRCh38
rs114389253 Chr.1: 150816399-150816399 on Build GRCh38
rs770675326 Chr.1: 150816329-150816329 on Build GRCh38
rs753587106 Chr.1: 150816278-150816278 on Build GRCh38
rs370635753 Chr.1: 150816427-150816427 on Build GRCh38

View Full Product Details