Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 1024878-1025137 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGFRL1
Entrez Gene ID
53834
Gene Name
fibroblast growth factor receptor-like 1
Gene Aliases
FGFR5, FHFR
UniGene
Hs.193326
Gene Chromosome Location
Chr.4: 1011822-1026898 on Build GRCh38
Transcripts
NM_001004356.2 NM_001004358.1 NM_021923.3 XM_011513486.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1049524 c.1273G>A p.D425N Chr.4: 1025105-1025105 on Build GRCh38
4684624 c.1038G>A p.P346P Chr.4: 1024960-1024960 on Build GRCh38
327801 c.1262G>A p.G421E Chr.4: 1025094-1025094 on Build GRCh38
1186498 c.1114G>T p.A372S Chr.4: 1024946-1024946 on Build GRCh38
327802 c.1172G>A p.G391E Chr.4: 1025094-1025094 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768252857 Chr.4: 1025046-1025046 on Build GRCh38
rs779615534 Chr.4: 1025029-1025029 on Build GRCh38
rs1000981403 Chr.4: 1025051-1025051 on Build GRCh38
rs750561767 Chr.4: 1025105-1025105 on Build GRCh38
rs577572432 Chr.4: 1025053-1025053 on Build GRCh38
rs760976677 Chr.4: 1025099-1025099 on Build GRCh38
rs907710217 Chr.4: 1025095-1025095 on Build GRCh38
rs368437012 Chr.4: 1025102-1025102 on Build GRCh38
rs752771234 Chr.4: 1024945-1024945 on Build GRCh38
rs200338999 Chr.4: 1025088-1025088 on Build GRCh38
rs373660136 Chr.4: 1024982-1024982 on Build GRCh38
rs746360727 Chr.4: 1025097-1025097 on Build GRCh38
rs778745736 Chr.4: 1024961-1024961 on Build GRCh38
rs753912109 Chr.4: 1024992-1024992 on Build GRCh38
rs747654089 Chr.4: 1025052-1025052 on Build GRCh38
rs771759616 Chr.4: 1024914-1024914 on Build GRCh38
rs759910460 Chr.4: 1025058-1025058 on Build GRCh38
rs375985073 Chr.4: 1025091-1025091 on Build GRCh38
rs146112374 Chr.4: 1024975-1024975 on Build GRCh38
rs943562921 Chr.4: 1025070-1025070 on Build GRCh38
rs745740200 Chr.4: 1024930-1024930 on Build GRCh38
rs368961378 Chr.4: 1024938-1024938 on Build GRCh38
rs376736117 Chr.4: 1025117-1025117 on Build GRCh38
rs374653229 Chr.4: 1025106-1025106 on Build GRCh38
rs567597911 Chr.4: 1024947-1024947 on Build GRCh38
rs201062727 Chr.4: 1025056-1025056 on Build GRCh38
rs911932019 Chr.4: 1025112-1025112 on Build GRCh38
rs774750247 Chr.4: 1024979-1024979 on Build GRCh38
rs755231134 Chr.4: 1024939-1024939 on Build GRCh38
rs369065988 Chr.4: 1024912-1024913 on Build GRCh38
rs370211317 Chr.4: 1025083-1025083 on Build GRCh38
rs767896163 Chr.4: 1024944-1024944 on Build GRCh38
rs752283163 Chr.4: 1025110-1025110 on Build GRCh38
rs61734883 Chr.4: 1025059-1025059 on Build GRCh38
rs201377348 Chr.4: 1025081-1025081 on Build GRCh38
rs773783296 Chr.4: 1024919-1024919 on Build GRCh38
rs202037959 Chr.4: 1024991-1024991 on Build GRCh38
rs537262686 Chr.4: 1024915-1024915 on Build GRCh38
rs749241683 Chr.4: 1024958-1024958 on Build GRCh38
rs750366202 Chr.4: 1025084-1025084 on Build GRCh38
rs763163157 Chr.4: 1024926-1024926 on Build GRCh38
rs745613727 Chr.4: 1025015-1025015 on Build GRCh38
rs375677549 Chr.4: 1024990-1024990 on Build GRCh38
rs759211786 Chr.4: 1024902-1024902 on Build GRCh38
rs776053533 Chr.4: 1025065-1025065 on Build GRCh38
rs777488330 Chr.4: 1025096-1025096 on Build GRCh38
rs776185476 Chr.4: 1025047-1025047 on Build GRCh38
rs939094385 Chr.4: 1025048-1025048 on Build GRCh38
rs371943795 Chr.4: 1024959-1024959 on Build GRCh38
rs959267006 Chr.4: 1025087-1025087 on Build GRCh38
rs4647930 Chr.4: 1024917-1024917 on Build GRCh38
rs766347101 Chr.4: 1025108-1025108 on Build GRCh38
rs775146211 Chr.4: 1024965-1024965 on Build GRCh38
rs758203364 Chr.4: 1025017-1025017 on Build GRCh38
rs756272863 Chr.4: 1024946-1024946 on Build GRCh38
rs140148549 Chr.4: 1024987-1024987 on Build GRCh38
rs746661915 Chr.4: 1025033-1025033 on Build GRCh38
rs764261493 Chr.4: 1025077-1025077 on Build GRCh38
rs754631527 Chr.4: 1025089-1025089 on Build GRCh38
rs760955260 Chr.4: 1025066-1025066 on Build GRCh38
rs925555901 Chr.4: 1025063-1025063 on Build GRCh38
rs111246053 Chr.4: 1024966-1024966 on Build GRCh38
rs778746987 Chr.4: 1025000-1025000 on Build GRCh38
rs757186468 Chr.4: 1024996-1024996 on Build GRCh38
rs375150331 Chr.4: 1025098-1025098 on Build GRCh38
rs376903367 Chr.4: 1024962-1024962 on Build GRCh38
rs4647931 Chr.4: 1025103-1025103 on Build GRCh38
rs766847951 Chr.4: 1024976-1024976 on Build GRCh38
rs890714676 Chr.4: 1025031-1025031 on Build GRCh38
rs367617448 Chr.4: 1025113-1025113 on Build GRCh38
rs774077336 Chr.4: 1024901-1024901 on Build GRCh38
rs765297271 Chr.4: 1025104-1025104 on Build GRCh38
rs751318362 Chr.4: 1025109-1025109 on Build GRCh38
rs201785648 Chr.4: 1025092-1025092 on Build GRCh38
rs200822222 Chr.4: 1025114-1025114 on Build GRCh38
rs997527849 Chr.4: 1024960-1024960 on Build GRCh38
rs373506035 Chr.4: 1025057-1025057 on Build GRCh38
rs962904426 Chr.4: 1025005-1025005 on Build GRCh38
rs201440347 Chr.4: 1024967-1024967 on Build GRCh38
rs758398284 Chr.4: 1025086-1025086 on Build GRCh38

View Full Product Details