Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 18256578-18256823 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHODL
Entrez Gene ID
140578
Gene Name
chondrolectin
Gene Aliases
C21orf68, MT75, PRED12
UniGene
Hs.283725
Gene Chromosome Location
Chr.21: 17819329-18267371 on Build GRCh38
Transcripts
NM_001204174.1 NM_001204175.1 NM_001204176.1 NM_001204177.1 NM_001204178.1 NM_024944.2 XM_011529453.2 XM_011529457.2 XM_017028273.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1615927 c.188G>T p.S63I Chr.21: 18256617-18256617 on Build GRCh38
5894108 c.269C>T p.P90L Chr.21: 18256698-18256698 on Build GRCh38
4100868 c.158G>T p.S53I Chr.21: 18256587-18256587 on Build GRCh38
1149388 c.138G>T p.L46L Chr.21: 18256690-18256690 on Build GRCh38
6004738 c.76G>T p.V26F Chr.21: 18256628-18256628 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756719942 Chr.21: 18256734-18256734 on Build GRCh38
rs755708329 Chr.21: 18256638-18256638 on Build GRCh38
rs61739664 Chr.21: 18256699-18256699 on Build GRCh38
rs976376931 Chr.21: 18256692-18256692 on Build GRCh38
rs532714129 Chr.21: 18256617-18256617 on Build GRCh38
rs920971021 Chr.21: 18256698-18256698 on Build GRCh38
rs142557853 Chr.21: 18256651-18256651 on Build GRCh38
rs749330837 Chr.21: 18256688-18256688 on Build GRCh38
rs751629840 Chr.21: 18256717-18256719 on Build GRCh38
rs767759893 Chr.21: 18256708-18256708 on Build GRCh38
rs781530660 Chr.21: 18256676-18256676 on Build GRCh38
rs777145979 Chr.21: 18256647-18256647 on Build GRCh38
rs759939160 Chr.21: 18256623-18256623 on Build GRCh38
rs758006539 Chr.21: 18256757-18256757 on Build GRCh38
rs148388759 Chr.21: 18256771-18256771 on Build GRCh38
rs549150011 Chr.21: 18256705-18256705 on Build GRCh38
rs370022813 Chr.21: 18256669-18256669 on Build GRCh38
rs373342182 Chr.21: 18256677-18256677 on Build GRCh38
rs760250647 Chr.21: 18256714-18256714 on Build GRCh38
rs375671735 Chr.21: 18256717-18256717 on Build GRCh38
rs775280248 Chr.21: 18256713-18256713 on Build GRCh38
rs377519818 Chr.21: 18256679-18256679 on Build GRCh38
rs540479831 Chr.21: 18256597-18256597 on Build GRCh38
rs147579421 Chr.21: 18256601-18256601 on Build GRCh38
rs773436892 Chr.21: 18256681-18256681 on Build GRCh38
rs769745802 Chr.21: 18256599-18256599 on Build GRCh38
rs753281252 Chr.21: 18256648-18256648 on Build GRCh38
rs752270530 Chr.21: 18256633-18256633 on Build GRCh38
rs200848816 Chr.21: 18256798-18256798 on Build GRCh38
rs567256236 Chr.21: 18256747-18256747 on Build GRCh38
rs746736900 Chr.21: 18256777-18256777 on Build GRCh38
rs375784288 Chr.21: 18256666-18256666 on Build GRCh38
rs753371867 Chr.21: 18256726-18256726 on Build GRCh38
rs778777155 Chr.21: 18256652-18256652 on Build GRCh38
rs765203113 Chr.21: 18256748-18256748 on Build GRCh38
rs751992493 Chr.21: 18256602-18256602 on Build GRCh38
rs146669901 Chr.21: 18256700-18256700 on Build GRCh38
rs749953036 Chr.21: 18256756-18256756 on Build GRCh38
rs144365214 Chr.21: 18256755-18256755 on Build GRCh38
rs745697776 Chr.21: 18256662-18256662 on Build GRCh38
rs756414774 Chr.21: 18256791-18256791 on Build GRCh38
rs767239738 Chr.21: 18256627-18256627 on Build GRCh38

View Full Product Details