Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6445037-6445262 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD27
Entrez Gene ID
939
Gene Name
CD27 molecule
Gene Aliases
S152, S152. LPFS2, T14, TNFRSF7, Tp55
UniGene
Hs.355307
Gene Chromosome Location
Chr.12: 6444867-6451718 on Build GRCh38
Transcripts
NM_001242.4 XM_017020232.1 XM_017020234.1 NR_015382.2

Gene Symbol
CD27-AS1
Entrez Gene ID
678655
Gene Name
CD27 antisense RNA 1
Gene Aliases
-
UniGene
Hs.655224
Gene Chromosome Location
Chr.12: 6439001-6451517 on Build GRCh38
Transcripts
NM_001242.4 XM_017020232.1 XM_017020234.1 NR_015382.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3700332 c.101C>A p.A34D Chr.12: 6445196-6445196 on Build GRCh38
4044173 c.9G>A p.R3R Chr.12: 6445104-6445104 on Build GRCh38
3987154 c.26T>A p.L9Q Chr.12: 6445121-6445121 on Build GRCh38
5948981 c.88A>G p.R30G Chr.12: 6445183-6445183 on Build GRCh38
3464254 c.17C>T p.P6L Chr.12: 6445112-6445112 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752132858 Chr.12: 6445086-6445086 on Build GRCh38
rs769004909 Chr.12: 6445133-6445133 on Build GRCh38
rs761928597 Chr.12: 6445166-6445166 on Build GRCh38
rs199640884 Chr.12: 6445081-6445081 on Build GRCh38
rs753465972 Chr.12: 6445241-6445241 on Build GRCh38
rs762885174 Chr.12: 6445179-6445179 on Build GRCh38
rs766179003 Chr.12: 6445186-6445186 on Build GRCh38
rs773972949 Chr.12: 6445200-6445200 on Build GRCh38
rs779787155 Chr.12: 6445179-6445180 on Build GRCh38
rs747125906 Chr.12: 6445237-6445238 on Build GRCh38
rs766298479 Chr.12: 6445065-6445065 on Build GRCh38
rs776856547 Chr.12: 6445149-6445149 on Build GRCh38
rs770201739 Chr.12: 6445169-6445169 on Build GRCh38
rs755775043 Chr.12: 6445102-6445102 on Build GRCh38
rs750165204 Chr.12: 6445125-6445128 on Build GRCh38
rs571145229 Chr.12: 6445124-6445124 on Build GRCh38
rs398122933 Chr.12: 6445119-6445119 on Build GRCh38
rs548323486 Chr.12: 6445061-6445061 on Build GRCh38
rs762832382 Chr.12: 6445064-6445064 on Build GRCh38
rs369839468 Chr.12: 6445201-6445201 on Build GRCh38
rs146726863 Chr.12: 6445132-6445132 on Build GRCh38
rs553686868 Chr.12: 6445158-6445158 on Build GRCh38
rs758426151 Chr.12: 6445162-6445163 on Build GRCh38
rs34540052 Chr.12: 6445125-6445125 on Build GRCh38
rs1032225550 Chr.12: 6445123-6445123 on Build GRCh38
rs767297512 Chr.12: 6445210-6445210 on Build GRCh38
rs200673347 Chr.12: 6445080-6445080 on Build GRCh38
rs1032752799 Chr.12: 6445209-6445209 on Build GRCh38
rs763774879 Chr.12: 6445236-6445236 on Build GRCh38
rs879501929 Chr.12: 6445220-6445220 on Build GRCh38
rs11569363 Chr.12: 6445066-6445066 on Build GRCh38
rs755615493 Chr.12: 6445230-6445230 on Build GRCh38
rs766034583 Chr.12: 6445223-6445223 on Build GRCh38
rs776469650 Chr.12: 6445103-6445103 on Build GRCh38
rs750260259 Chr.12: 6445060-6445060 on Build GRCh38
rs752259578 Chr.12: 6445217-6445217 on Build GRCh38
rs772449872 Chr.12: 6445126-6445126 on Build GRCh38
rs773567512 Chr.12: 6445172-6445172 on Build GRCh38
rs975388482 Chr.12: 6445162-6445162 on Build GRCh38

View Full Product Details