Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 39144258-39144505 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSRNP1
Entrez Gene ID
64651
Gene Name
cysteine and serine rich nuclear protein 1
Gene Aliases
AXUD1, CSRNP-1, FAM130B, TAIP-3, URAX1
UniGene
Hs.370950
Gene Chromosome Location
Chr.3: 39141851-39154602 on Build GRCh38
Transcripts
NM_001320559.1 NM_001320560.1 NM_033027.3 XM_017007048.1 XM_017007049.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5780337 c.657C>A p.I219I Chr.3: 39144260-39144260 on Build GRCh38
4917203 c.622C>A p.R208R Chr.3: 39144295-39144295 on Build GRCh38
446477 c.657C>T p.I219I Chr.3: 39144260-39144260 on Build GRCh38
1044465 c.623G>A p.R208Q Chr.3: 39144294-39144294 on Build GRCh38
3593798 c.583G>A p.E195K Chr.3: 39144334-39144334 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138372931 Chr.3: 39144365-39144365 on Build GRCh38
rs758544588 Chr.3: 39144344-39144344 on Build GRCh38
rs890879591 Chr.3: 39144414-39144414 on Build GRCh38
rs781483967 Chr.3: 39144447-39144447 on Build GRCh38
rs777108852 Chr.3: 39144312-39144312 on Build GRCh38
rs765922302 Chr.3: 39144334-39144334 on Build GRCh38
rs770181827 Chr.3: 39144454-39144454 on Build GRCh38
rs748157430 Chr.3: 39144280-39144280 on Build GRCh38
rs941546443 Chr.3: 39144330-39144330 on Build GRCh38
rs568323801 Chr.3: 39144466-39144466 on Build GRCh38
rs759270599 Chr.3: 39144385-39144385 on Build GRCh38
rs1041674662 Chr.3: 39144399-39144399 on Build GRCh38
rs776980215 Chr.3: 39144358-39144358 on Build GRCh38
rs775832050 Chr.3: 39144303-39144303 on Build GRCh38
rs745915113 Chr.3: 39144343-39144343 on Build GRCh38
rs762321878 Chr.3: 39144323-39144323 on Build GRCh38
rs752619572 Chr.3: 39144340-39144340 on Build GRCh38
rs755219744 Chr.3: 39144446-39144446 on Build GRCh38
rs779011156 Chr.3: 39144461-39144461 on Build GRCh38
rs199818998 Chr.3: 39144464-39144464 on Build GRCh38
rs910032303 Chr.3: 39144319-39144319 on Build GRCh38
rs779138699 Chr.3: 39144298-39144298 on Build GRCh38
rs745818618 Chr.3: 39144369-39144371 on Build GRCh38
rs540112925 Chr.3: 39144309-39144309 on Build GRCh38
rs765975625 Chr.3: 39144384-39144384 on Build GRCh38
rs772625731 Chr.3: 39144329-39144329 on Build GRCh38
rs778131994 Chr.3: 39144460-39144460 on Build GRCh38
rs764656490 Chr.3: 39144326-39144326 on Build GRCh38
rs757359136 Chr.3: 39144294-39144294 on Build GRCh38
rs200004556 Chr.3: 39144347-39144347 on Build GRCh38
rs754810673 Chr.3: 39144339-39144339 on Build GRCh38
rs749818099 Chr.3: 39144463-39144463 on Build GRCh38
rs375176179 Chr.3: 39144304-39144304 on Build GRCh38
rs762367732 Chr.3: 39144381-39144381 on Build GRCh38
rs562658217 Chr.3: 39144386-39144386 on Build GRCh38
rs574676260 Chr.3: 39144292-39144292 on Build GRCh38
rs560006152 Chr.3: 39144331-39144331 on Build GRCh38
rs145888015 Chr.3: 39144409-39144409 on Build GRCh38
rs752562774 Chr.3: 39144388-39144388 on Build GRCh38
rs753878701 Chr.3: 39144412-39144412 on Build GRCh38
rs775323926 Chr.3: 39144469-39144469 on Build GRCh38
rs927189569 Chr.3: 39144361-39144361 on Build GRCh38
rs772207810 Chr.3: 39144302-39144302 on Build GRCh38
rs773816035 Chr.3: 39144379-39144379 on Build GRCh38
rs810743 Chr.3: 39144477-39144477 on Build GRCh38
rs749571779 Chr.3: 39144348-39144348 on Build GRCh38
rs751294413 Chr.3: 39144337-39144337 on Build GRCh38
rs773946995 Chr.3: 39144465-39144465 on Build GRCh38
rs758393111 Chr.3: 39144435-39144435 on Build GRCh38
rs746064037 Chr.3: 39144300-39144300 on Build GRCh38
rs780194452 Chr.3: 39144441-39144441 on Build GRCh38
rs764006328 Chr.3: 39144476-39144476 on Build GRCh38
rs112352444 Chr.3: 39144442-39144442 on Build GRCh38
rs748487285 Chr.3: 39144452-39144452 on Build GRCh38
rs768950919 Chr.3: 39144355-39144355 on Build GRCh38
rs761814951 Chr.3: 39144481-39144481 on Build GRCh38
rs760540295 Chr.3: 39144474-39144474 on Build GRCh38
rs532218961 Chr.3: 39144342-39144342 on Build GRCh38
rs531666797 Chr.3: 39144406-39144406 on Build GRCh38

View Full Product Details