Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 81992813-81993056 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPATA31D1
Entrez Gene ID
389763
Gene Name
SPATA31 subfamily D member 1
Gene Aliases
FAM75D1
UniGene
Hs.209545
Gene Chromosome Location
Chr.9: 81987404-81995256 on Build GRCh38
Transcripts
NM_001001670.2 XM_017014710.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1110484 c.2562G>A p.W854* Chr.9: 81993032-81993032 on Build GRCh38
1110479 c.2366C>T p.P789L Chr.9: 81992836-81992836 on Build GRCh38
5661013 c.2583G>A p.M861I Chr.9: 81993053-81993053 on Build GRCh38
4139609 c.2363G>T p.G788V Chr.9: 81992833-81992833 on Build GRCh38
1110483 c.2562G>A p.W854* Chr.9: 81993032-81993032 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372805132 Chr.9: 81993004-81993004 on Build GRCh38
rs771650425 Chr.9: 81992899-81992899 on Build GRCh38
rs751339154 Chr.9: 81992921-81992922 on Build GRCh38
rs767801695 Chr.9: 81993018-81993018 on Build GRCh38
rs751718454 Chr.9: 81992935-81992935 on Build GRCh38
rs199711694 Chr.9: 81992907-81992907 on Build GRCh38
rs201491537 Chr.9: 81992870-81992870 on Build GRCh38
rs568626063 Chr.9: 81992988-81992988 on Build GRCh38
rs372310314 Chr.9: 81992968-81992968 on Build GRCh38
rs759725073 Chr.9: 81993013-81993013 on Build GRCh38
rs763574014 Chr.9: 81993001-81993001 on Build GRCh38
rs1004480271 Chr.9: 81992863-81992863 on Build GRCh38
rs992271271 Chr.9: 81992887-81992887 on Build GRCh38
rs757110653 Chr.9: 81993021-81993021 on Build GRCh38
rs749112869 Chr.9: 81992868-81992868 on Build GRCh38
rs533906009 Chr.9: 81992904-81992904 on Build GRCh38
rs556717382 Chr.9: 81992973-81992973 on Build GRCh38
rs770185961 Chr.9: 81992995-81992995 on Build GRCh38
rs766549142 Chr.9: 81992931-81992931 on Build GRCh38
rs755857922 Chr.9: 81992865-81992865 on Build GRCh38
rs778974980 Chr.9: 81992958-81992958 on Build GRCh38
rs376973200 Chr.9: 81992854-81992854 on Build GRCh38
rs765682060 Chr.9: 81992918-81992918 on Build GRCh38
rs1052364388 Chr.9: 81992983-81992983 on Build GRCh38
rs771466098 Chr.9: 81992961-81992961 on Build GRCh38
rs773945588 Chr.9: 81992836-81992836 on Build GRCh38
rs774998083 Chr.9: 81992900-81992900 on Build GRCh38
rs745324500 Chr.9: 81992893-81992893 on Build GRCh38
rs369693916 Chr.9: 81992853-81992853 on Build GRCh38
rs750986595 Chr.9: 81992925-81992925 on Build GRCh38
rs374373990 Chr.9: 81992937-81992937 on Build GRCh38
rs375606595 Chr.9: 81993030-81993030 on Build GRCh38
rs760429504 Chr.9: 81992901-81992901 on Build GRCh38
rs780119147 Chr.9: 81992862-81992862 on Build GRCh38
rs913772836 Chr.9: 81992996-81992996 on Build GRCh38
rs1027176653 Chr.9: 81992852-81992852 on Build GRCh38
rs778823280 Chr.9: 81993028-81993028 on Build GRCh38
rs764583739 Chr.9: 81992845-81992845 on Build GRCh38
rs762195403 Chr.9: 81992912-81992912 on Build GRCh38
rs566544985 Chr.9: 81992881-81992881 on Build GRCh38
rs775232690 Chr.9: 81992841-81992841 on Build GRCh38
rs758526262 Chr.9: 81992857-81992857 on Build GRCh38
rs370326842 Chr.9: 81992866-81992866 on Build GRCh38
rs750462192 Chr.9: 81992855-81992855 on Build GRCh38
rs762913197 Chr.9: 81992930-81992930 on Build GRCh38
rs374062634 Chr.9: 81992846-81992846 on Build GRCh38
rs372215050 Chr.9: 81992837-81992837 on Build GRCh38
rs747297583 Chr.9: 81992864-81992864 on Build GRCh38
rs764005264 Chr.9: 81992893-81992897 on Build GRCh38
rs552420674 Chr.9: 81992946-81992946 on Build GRCh38
rs372028836 Chr.9: 81992967-81992967 on Build GRCh38
rs773588800 Chr.9: 81992997-81992997 on Build GRCh38
rs757143269 Chr.9: 81992947-81992947 on Build GRCh38
rs10867826 Chr.9: 81992843-81992843 on Build GRCh38

View Full Product Details