Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 203698120-203698385 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP2B4
Entrez Gene ID
493
Gene Name
ATPase plasma membrane Ca2+ transporting 4
Gene Aliases
ATP2B2, MXRA1, PMCA4, PMCA4b, PMCA4x
UniGene
Hs.343522
Gene Chromosome Location
Chr.1: 203626787-203744081 on Build GRCh38
Transcripts
NM_001001396.2 NM_001684.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1639690 c.230G>T p.R77M Chr.1: 203698193-203698193 on Build GRCh38
902491 c.295G>T p.E99* Chr.1: 203698258-203698258 on Build GRCh38
3864160 c.325C>T p.L109L Chr.1: 203698288-203698288 on Build GRCh38
902489 c.283G>T p.E95* Chr.1: 203698246-203698246 on Build GRCh38
6607807 c.260C>A p.P87H Chr.1: 203698223-203698223 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144377803 Chr.1: 203698327-203698327 on Build GRCh38
rs143571623 Chr.1: 203698328-203698328 on Build GRCh38
rs2228445 Chr.1: 203698281-203698281 on Build GRCh38
rs138303385 Chr.1: 203698222-203698222 on Build GRCh38
rs527399964 Chr.1: 203698278-203698278 on Build GRCh38
rs201177984 Chr.1: 203698221-203698221 on Build GRCh38
rs975041778 Chr.1: 203698229-203698229 on Build GRCh38
rs771816695 Chr.1: 203698223-203698223 on Build GRCh38
rs779601653 Chr.1: 203698330-203698330 on Build GRCh38
rs760570158 Chr.1: 203698164-203698164 on Build GRCh38
rs756449111 Chr.1: 203698189-203698189 on Build GRCh38
rs769726180 Chr.1: 203698217-203698218 on Build GRCh38
rs777154265 Chr.1: 203698354-203698354 on Build GRCh38
rs753769605 Chr.1: 203698171-203698171 on Build GRCh38
rs751699115 Chr.1: 203698182-203698182 on Build GRCh38
rs766708513 Chr.1: 203698305-203698305 on Build GRCh38
rs759460032 Chr.1: 203698238-203698238 on Build GRCh38
rs750729824 Chr.1: 203698184-203698184 on Build GRCh38
rs776307408 Chr.1: 203698237-203698237 on Build GRCh38
rs766110790 Chr.1: 203698166-203698166 on Build GRCh38
rs562320431 Chr.1: 203698190-203698190 on Build GRCh38
rs750615512 Chr.1: 203698282-203698282 on Build GRCh38
rs376176284 Chr.1: 203698308-203698308 on Build GRCh38
rs755555037 Chr.1: 203698213-203698213 on Build GRCh38
rs200395561 Chr.1: 203698224-203698224 on Build GRCh38
rs114358697 Chr.1: 203698331-203698331 on Build GRCh38
rs765229515 Chr.1: 203698258-203698258 on Build GRCh38
rs764106247 Chr.1: 203698180-203698180 on Build GRCh38
rs756290397 Chr.1: 203698290-203698290 on Build GRCh38
rs531572158 Chr.1: 203698212-203698212 on Build GRCh38
rs774386259 Chr.1: 203698266-203698266 on Build GRCh38
rs372768311 Chr.1: 203698339-203698339 on Build GRCh38
rs542534953 Chr.1: 203698183-203698183 on Build GRCh38
rs759474365 Chr.1: 203698177-203698177 on Build GRCh38
rs772877126 Chr.1: 203698160-203698160 on Build GRCh38

View Full Product Details