Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 57415734-57415981 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCL17
Entrez Gene ID
6361
Gene Name
C-C motif chemokine ligand 17
Gene Aliases
A-152E5.3, ABCD-2, SCYA17, TARC
UniGene
Hs.546294
Gene Chromosome Location
Chr.16: 57396076-57416063 on Build GRCh38
Transcripts
NM_002987.2 XM_011523256.2 XM_017023530.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
214301 c.206G>A p.G69D Chr.16: 57415782-57415782 on Build GRCh38
3387503 c.205G>A p.G69S Chr.16: 57415781-57415781 on Build GRCh38
971827 c.269G>T p.S90I Chr.16: 57415845-57415845 on Build GRCh38
4484522 c.281C>T p.S94F Chr.16: 57415857-57415857 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs529747644 Chr.16: 57415754-57415756 on Build GRCh38
rs765518571 Chr.16: 57415844-57415844 on Build GRCh38
rs369498202 Chr.16: 57415880-57415880 on Build GRCh38
rs34379253 Chr.16: 57415775-57415775 on Build GRCh38
rs984737757 Chr.16: 57415756-57415756 on Build GRCh38
rs756855167 Chr.16: 57415886-57415887 on Build GRCh38
rs748381602 Chr.16: 57415776-57415776 on Build GRCh38
rs561153052 Chr.16: 57415957-57415957 on Build GRCh38
rs946310208 Chr.16: 57415924-57415924 on Build GRCh38
rs748319436 Chr.16: 57415804-57415806 on Build GRCh38
rs936294432 Chr.16: 57415888-57415888 on Build GRCh38
rs201634344 Chr.16: 57415807-57415807 on Build GRCh38
rs570762119 Chr.16: 57415781-57415781 on Build GRCh38
rs377119374 Chr.16: 57415873-57415873 on Build GRCh38
rs542999826 Chr.16: 57415945-57415945 on Build GRCh38
rs754957215 Chr.16: 57415895-57415895 on Build GRCh38
rs901979152 Chr.16: 57415944-57415944 on Build GRCh38
rs753101348 Chr.16: 57415867-57415867 on Build GRCh38
rs778905686 Chr.16: 57415913-57415913 on Build GRCh38
rs372670099 Chr.16: 57415797-57415797 on Build GRCh38
rs559175146 Chr.16: 57415878-57415878 on Build GRCh38
rs149709463 Chr.16: 57415798-57415798 on Build GRCh38
rs766559717 Chr.16: 57415922-57415922 on Build GRCh38
rs201176349 Chr.16: 57415894-57415894 on Build GRCh38
rs777076468 Chr.16: 57415811-57415811 on Build GRCh38
rs748859619 Chr.16: 57415870-57415871 on Build GRCh38
rs768510723 Chr.16: 57415770-57415770 on Build GRCh38
rs759909616 Chr.16: 57415815-57415815 on Build GRCh38
rs78838340 Chr.16: 57415959-57415959 on Build GRCh38
rs947826098 Chr.16: 57415864-57415864 on Build GRCh38
rs753984507 Chr.16: 57415929-57415929 on Build GRCh38

View Full Product Details