Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 74190130-74190387 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF4H
Entrez Gene ID
7458
Gene Name
eukaryotic translation initiation factor 4H
Gene Aliases
WBSCR1, WSCR1, eIF-4H
UniGene
Hs.520943
Gene Chromosome Location
Chr.7: 74174376-74197101 on Build GRCh38
Transcripts
NM_022170.1 NM_031992.1 XM_011516556.1 XM_011516557.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7104392 c.469+1G>A p.? Chr.7: 74190307-74190307 on Build GRCh38
4149336 c.424C>T p.R142* Chr.7: 74190261-74190261 on Build GRCh38
5035928 c.415G>A p.G139S Chr.7: 74190252-74190252 on Build GRCh38
7133541 c.431C>T p.S144F Chr.7: 74190268-74190268 on Build GRCh38
5935264 c.410-6C>T p.? Chr.7: 74190241-74190241 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1028680528 Chr.7: 74190175-74190175 on Build GRCh38
rs781892481 Chr.7: 74190338-74190338 on Build GRCh38
rs782693125 Chr.7: 74190358-74190358 on Build GRCh38
rs781930395 Chr.7: 74190335-74190335 on Build GRCh38
rs782389714 Chr.7: 74190306-74190306 on Build GRCh38
rs782674081 Chr.7: 74190257-74190257 on Build GRCh38
rs782214719 Chr.7: 74190288-74190288 on Build GRCh38
rs781799285 Chr.7: 74190241-74190241 on Build GRCh38
rs781841187 Chr.7: 74190348-74190348 on Build GRCh38
rs781799890 Chr.7: 74190235-74190235 on Build GRCh38
rs782061476 Chr.7: 74190354-74190355 on Build GRCh38
rs782111617 Chr.7: 74190196-74190196 on Build GRCh38
rs369255921 Chr.7: 74190313-74190313 on Build GRCh38
rs781946662 Chr.7: 74190341-74190341 on Build GRCh38
rs782380321 Chr.7: 74190276-74190276 on Build GRCh38
rs782466812 Chr.7: 74190349-74190349 on Build GRCh38
rs782052937 Chr.7: 74190222-74190223 on Build GRCh38
rs981332285 Chr.7: 74190268-74190268 on Build GRCh38
rs781980423 Chr.7: 74190309-74190309 on Build GRCh38
rs782610397 Chr.7: 74190287-74190287 on Build GRCh38
rs142964239 Chr.7: 74190166-74190166 on Build GRCh38
rs61732247 Chr.7: 74190289-74190289 on Build GRCh38
rs782757051 Chr.7: 74190318-74190319 on Build GRCh38
rs782109541 Chr.7: 74190350-74190350 on Build GRCh38
rs782736746 Chr.7: 74190321-74190321 on Build GRCh38
rs368681152 Chr.7: 74190229-74190229 on Build GRCh38
rs782362543 Chr.7: 74190197-74190197 on Build GRCh38
rs782788335 Chr.7: 74190199-74190199 on Build GRCh38
rs781890890 Chr.7: 74190214-74190214 on Build GRCh38
rs373771109 Chr.7: 74190301-74190301 on Build GRCh38
rs782132905 Chr.7: 74190222-74190222 on Build GRCh38
rs782808583 Chr.7: 74190337-74190337 on Build GRCh38
rs782033235 Chr.7: 74190296-74190296 on Build GRCh38
rs782271175 Chr.7: 74190261-74190261 on Build GRCh38
rs782058897 Chr.7: 74190362-74190362 on Build GRCh38
rs782405532 Chr.7: 74190228-74190229 on Build GRCh38
rs782094367 Chr.7: 74190312-74190312 on Build GRCh38
rs377427598 Chr.7: 74190211-74190211 on Build GRCh38
rs143662860 Chr.7: 74190249-74190249 on Build GRCh38
rs782807691 Chr.7: 74190213-74190213 on Build GRCh38
rs781831841 Chr.7: 74190228-74190228 on Build GRCh38
rs377167722 Chr.7: 74190344-74190344 on Build GRCh38

View Full Product Details