Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 68866181-68866406 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMP10
Entrez Gene ID
27302
Gene Name
bone morphogenetic protein 10
Gene Aliases
-
UniGene
Hs.158317
Gene Chromosome Location
Chr.2: 68865481-68871517 on Build GRCh38
Transcripts
NM_014482.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3055877 c.584A>C p.N195T Chr.2: 68866322-68866322 on Build GRCh38
1022384 c.542G>T p.R181I Chr.2: 68866364-68866364 on Build GRCh38
1022383 c.571A>G p.I191V Chr.2: 68866335-68866335 on Build GRCh38
3939136 c.712C>T p.R238W Chr.2: 68866194-68866194 on Build GRCh38
6010138 c.664G>A p.V222I Chr.2: 68866242-68866242 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769205782 Chr.2: 68866321-68866321 on Build GRCh38
rs769419535 Chr.2: 68866221-68866221 on Build GRCh38
rs748857725 Chr.2: 68866315-68866315 on Build GRCh38
rs372547313 Chr.2: 68866257-68866257 on Build GRCh38
rs529530758 Chr.2: 68866338-68866338 on Build GRCh38
rs746919370 Chr.2: 68866202-68866202 on Build GRCh38
rs780683420 Chr.2: 68866289-68866289 on Build GRCh38
rs774802365 Chr.2: 68866322-68866322 on Build GRCh38
rs376786943 Chr.2: 68866280-68866280 on Build GRCh38
rs775254780 Chr.2: 68866311-68866311 on Build GRCh38
rs757128148 Chr.2: 68866286-68866286 on Build GRCh38
rs769352228 Chr.2: 68866309-68866309 on Build GRCh38
rs758254088 Chr.2: 68866268-68866268 on Build GRCh38
rs763324940 Chr.2: 68866228-68866228 on Build GRCh38
rs750453264 Chr.2: 68866236-68866236 on Build GRCh38
rs757216135 Chr.2: 68866378-68866378 on Build GRCh38
rs369259498 Chr.2: 68866231-68866231 on Build GRCh38
rs759741960 Chr.2: 68866356-68866356 on Build GRCh38
rs767721958 Chr.2: 68866234-68866234 on Build GRCh38
rs766575868 Chr.2: 68866254-68866254 on Build GRCh38
rs982448276 Chr.2: 68866283-68866283 on Build GRCh38
rs1051238362 Chr.2: 68866364-68866364 on Build GRCh38
rs2231343 Chr.2: 68866222-68866222 on Build GRCh38
rs899557120 Chr.2: 68866358-68866358 on Build GRCh38
rs756324872 Chr.2: 68866243-68866243 on Build GRCh38
rs745520256 Chr.2: 68866219-68866219 on Build GRCh38
rs375225348 Chr.2: 68866229-68866229 on Build GRCh38
rs781057330 Chr.2: 68866380-68866380 on Build GRCh38
rs762419037 Chr.2: 68866327-68866327 on Build GRCh38
rs371768311 Chr.2: 68866213-68866213 on Build GRCh38
rs34008398 Chr.2: 68866281-68866281 on Build GRCh38
rs763889510 Chr.2: 68866359-68866359 on Build GRCh38
rs751287726 Chr.2: 68866374-68866374 on Build GRCh38
rs760859777 Chr.2: 68866340-68866340 on Build GRCh38
rs773189718 Chr.2: 68866339-68866339 on Build GRCh38
rs754157716 Chr.2: 68866353-68866353 on Build GRCh38
rs369791897 Chr.2: 68866350-68866350 on Build GRCh38
rs770494705 Chr.2: 68866209-68866209 on Build GRCh38
rs776105366 Chr.2: 68866214-68866214 on Build GRCh38
rs2231342 Chr.2: 68866307-68866307 on Build GRCh38
rs755687639 Chr.2: 68866383-68866383 on Build GRCh38

View Full Product Details