Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 90878929-90879158 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FURIN
Entrez Gene ID
5045
Gene Name
furin, paired basic amino acid cleaving enzyme
Gene Aliases
FUR, PACE, PCSK3, SPC1
UniGene
Hs.513153
Gene Chromosome Location
Chr.15: 90868592-90883458 on Build GRCh38
Transcripts
NM_001289823.1 NM_001289824.1 NM_002569.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4775600 c.1016C>T p.T339M Chr.15: 90878939-90878939 on Build GRCh38
4057918 c.1028G>A p.S343N Chr.15: 90878951-90878951 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs996978784 Chr.15: 90879059-90879059 on Build GRCh38
rs1012744453 Chr.15: 90879108-90879108 on Build GRCh38
rs917418936 Chr.15: 90878968-90878968 on Build GRCh38
rs765195561 Chr.15: 90878949-90878949 on Build GRCh38
rs754552007 Chr.15: 90879005-90879005 on Build GRCh38
rs779701983 Chr.15: 90878991-90878991 on Build GRCh38
rs560035806 Chr.15: 90879095-90879095 on Build GRCh38
rs149143094 Chr.15: 90878976-90878976 on Build GRCh38
rs557382838 Chr.15: 90879096-90879096 on Build GRCh38
rs28634912 Chr.15: 90879078-90879078 on Build GRCh38
rs1038401209 Chr.15: 90879045-90879045 on Build GRCh38
rs908515116 Chr.15: 90879021-90879021 on Build GRCh38
rs750221222 Chr.15: 90878983-90878983 on Build GRCh38
rs1044553971 Chr.15: 90879115-90879115 on Build GRCh38
rs541697573 Chr.15: 90879014-90879014 on Build GRCh38
rs941339763 Chr.15: 90879035-90879035 on Build GRCh38
rs899904527 Chr.15: 90879058-90879058 on Build GRCh38
rs1050873951 Chr.15: 90879070-90879070 on Build GRCh38
rs755844342 Chr.15: 90878988-90878988 on Build GRCh38
rs778260657 Chr.15: 90879011-90879011 on Build GRCh38
rs574808270 Chr.15: 90878996-90878996 on Build GRCh38
rs151289535 Chr.15: 90879096-90879096 on Build GRCh38
rs1057062879 Chr.15: 90879091-90879091 on Build GRCh38
rs1020866271 Chr.15: 90879106-90879106 on Build GRCh38
rs8030001 Chr.15: 90879103-90879103 on Build GRCh38
rs12438243 Chr.15: 90879094-90879094 on Build GRCh38
rs751556003 Chr.15: 90878955-90878955 on Build GRCh38
rs968023663 Chr.15: 90879122-90879122 on Build GRCh38

View Full Product Details