Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 47142043-47142302 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP4A22
Entrez Gene ID
284541
Gene Name
cytochrome P450 family 4 subfamily A member 22
Gene Aliases
-
UniGene
Hs.726474
Gene Chromosome Location
Chr.1: 47137425-47149738 on Build GRCh38
Transcripts
NM_001010969.3 NM_001308102.1 XM_005270767.3 XM_005270770.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5043340 c.428G>T p.R143L Chr.1: 47142153-47142153 on Build GRCh38
910393 c.412A>G p.T138A Chr.1: 47142137-47142137 on Build GRCh38
3751258 c.388G>A p.G130S Chr.1: 47142113-47142113 on Build GRCh38
122467 c.428G>A p.R143Q Chr.1: 47142153-47142153 on Build GRCh38
6639337 c.427C>T p.R143* Chr.1: 47142152-47142152 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2758715 Chr.1: 47142199-47142199 on Build GRCh38
rs2056899 Chr.1: 47142179-47142179 on Build GRCh38
rs138940178 Chr.1: 47142153-47142153 on Build GRCh38
rs753825027 Chr.1: 47142114-47142114 on Build GRCh38
rs1021073828 Chr.1: 47142220-47142221 on Build GRCh38
rs753898677 Chr.1: 47142067-47142067 on Build GRCh38
rs781758484 Chr.1: 47142156-47142156 on Build GRCh38
rs759406647 Chr.1: 47142198-47142198 on Build GRCh38
rs760443382 Chr.1: 47142184-47142184 on Build GRCh38
rs778862320 Chr.1: 47142236-47142239 on Build GRCh38
rs369859955 Chr.1: 47142074-47142074 on Build GRCh38
rs377315324 Chr.1: 47142103-47142103 on Build GRCh38
rs771740217 Chr.1: 47142180-47142180 on Build GRCh38
rs1049441295 Chr.1: 47142142-47142142 on Build GRCh38
rs770666322 Chr.1: 47142187-47142187 on Build GRCh38
rs71645850 Chr.1: 47142107-47142107 on Build GRCh38
rs376345723 Chr.1: 47142207-47142207 on Build GRCh38
rs755093071 Chr.1: 47142070-47142070 on Build GRCh38
rs200279801 Chr.1: 47142182-47142182 on Build GRCh38
rs367677710 Chr.1: 47142261-47142261 on Build GRCh38
rs752724599 Chr.1: 47142136-47142136 on Build GRCh38
rs555070132 Chr.1: 47142088-47142088 on Build GRCh38
rs747687099 Chr.1: 47142264-47142264 on Build GRCh38
rs757574338 Chr.1: 47142204-47142204 on Build GRCh38
rs751777511 Chr.1: 47142201-47142201 on Build GRCh38
rs747347393 Chr.1: 47142093-47142093 on Build GRCh38
rs756628501 Chr.1: 47142252-47142252 on Build GRCh38
rs763011921 Chr.1: 47142200-47142200 on Build GRCh38
rs759496666 Chr.1: 47142118-47142118 on Build GRCh38
rs778416551 Chr.1: 47142165-47142165 on Build GRCh38
rs766314584 Chr.1: 47142065-47142065 on Build GRCh38
rs776446557 Chr.1: 47142193-47142193 on Build GRCh38
rs371221965 Chr.1: 47142140-47142140 on Build GRCh38
rs925597036 Chr.1: 47142086-47142086 on Build GRCh38
rs757897972 Chr.1: 47142268-47142268 on Build GRCh38
rs775105535 Chr.1: 47142095-47142095 on Build GRCh38
rs771623586 Chr.1: 47142106-47142106 on Build GRCh38
rs2056901 Chr.1: 47142090-47142090 on Build GRCh38
rs897652439 Chr.1: 47142116-47142116 on Build GRCh38
rs768329252 Chr.1: 47142102-47142102 on Build GRCh38
rs746327819 Chr.1: 47142158-47142158 on Build GRCh38
rs748774551 Chr.1: 47142096-47142096 on Build GRCh38
rs201496093 Chr.1: 47142225-47142225 on Build GRCh38
rs772997965 Chr.1: 47142112-47142112 on Build GRCh38
rs758588354 Chr.1: 47142138-47142138 on Build GRCh38
rs147010308 Chr.1: 47142155-47142155 on Build GRCh38
rs771031011 Chr.1: 47142224-47142224 on Build GRCh38
rs771552978 Chr.1: 47142094-47142094 on Build GRCh38
rs777367506 Chr.1: 47142277-47142277 on Build GRCh38
rs189269010 Chr.1: 47142211-47142211 on Build GRCh38
rs779087879 Chr.1: 47142072-47142072 on Build GRCh38

View Full Product Details