Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 51456107-51456332 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMXL2
Entrez Gene ID
23312
Gene Name
Dmx like 2
Gene Aliases
PEPNS, RC3
UniGene
Hs.511386
Gene Chromosome Location
Chr.15: 51447724-51622833 on Build GRCh38
Transcripts
NM_001174116.1 NM_001174117.1 NM_015263.3 XM_005254255.1 XM_005254256.1 XM_017022034.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5920807 c.8384C>T p.P2795L Chr.15: 51456142-51456142 on Build GRCh38
239589 c.8341G>T p.G2781C Chr.15: 51456185-51456185 on Build GRCh38
4441643 c.8353G>A p.G2785S Chr.15: 51456173-51456173 on Build GRCh38
6479043 c.8344G>A p.A2782T Chr.15: 51456182-51456182 on Build GRCh38
7273715 c.8325T>A p.H2775Q Chr.15: 51456316-51456316 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144407842 Chr.15: 51456293-51456293 on Build GRCh38
rs775760774 Chr.15: 51456225-51456229 on Build GRCh38
rs949926120 Chr.15: 51456257-51456257 on Build GRCh38
rs779186222 Chr.15: 51456228-51456228 on Build GRCh38
rs772517464 Chr.15: 51456149-51456149 on Build GRCh38
rs777131630 Chr.15: 51456282-51456283 on Build GRCh38
rs769856719 Chr.15: 51456145-51456145 on Build GRCh38
rs200088879 Chr.15: 51456284-51456284 on Build GRCh38
rs761288719 Chr.15: 51456264-51456264 on Build GRCh38
rs763675377 Chr.15: 51456164-51456164 on Build GRCh38
rs771139832 Chr.15: 51456203-51456204 on Build GRCh38
rs776064763 Chr.15: 51456155-51456155 on Build GRCh38
rs527513265 Chr.15: 51456278-51456278 on Build GRCh38
rs369687812 Chr.15: 51456138-51456138 on Build GRCh38
rs373337134 Chr.15: 51456174-51456174 on Build GRCh38
rs971197899 Chr.15: 51456144-51456144 on Build GRCh38
rs559890629 Chr.15: 51456208-51456208 on Build GRCh38
rs769099788 Chr.15: 51456246-51456250 on Build GRCh38
rs764855578 Chr.15: 51456281-51456281 on Build GRCh38
rs1054418131 Chr.15: 51456180-51456180 on Build GRCh38
rs772451700 Chr.15: 51456241-51456241 on Build GRCh38
rs370064925 Chr.15: 51456201-51456201 on Build GRCh38
rs372985007 Chr.15: 51456200-51456200 on Build GRCh38
rs759889759 Chr.15: 51456148-51456148 on Build GRCh38
rs917085479 Chr.15: 51456250-51456250 on Build GRCh38
rs749487323 Chr.15: 51456146-51456146 on Build GRCh38
rs141041082 Chr.15: 51456147-51456147 on Build GRCh38
rs750085308 Chr.15: 51456292-51456292 on Build GRCh38
rs557865159 Chr.15: 51456289-51456290 on Build GRCh38
rs555921038 Chr.15: 51456207-51456207 on Build GRCh38
rs751463582 Chr.15: 51456202-51456202 on Build GRCh38
rs1051017287 Chr.15: 51456294-51456294 on Build GRCh38
rs539588036 Chr.15: 51456272-51456272 on Build GRCh38
rs984805593 Chr.15: 51456182-51456182 on Build GRCh38
rs377627316 Chr.15: 51456135-51456135 on Build GRCh38
rs771073694 Chr.15: 51456219-51456219 on Build GRCh38
rs750197901 Chr.15: 51456186-51456186 on Build GRCh38
rs772463836 Chr.15: 51456218-51456219 on Build GRCh38
rs749324171 Chr.15: 51456216-51456216 on Build GRCh38
rs746290295 Chr.15: 51456235-51456235 on Build GRCh38
rs753351209 Chr.15: 51456170-51456170 on Build GRCh38
rs751303807 Chr.15: 51456300-51456300 on Build GRCh38
rs551131295 Chr.15: 51456204-51456204 on Build GRCh38
rs761591199 Chr.15: 51456171-51456171 on Build GRCh38
rs761136653 Chr.15: 51456230-51456231 on Build GRCh38
rs776283339 Chr.15: 51456276-51456276 on Build GRCh38
rs200181576 Chr.15: 51456210-51456211 on Build GRCh38
rs758198930 Chr.15: 51456198-51456198 on Build GRCh38
rs776189127 Chr.15: 51456259-51456259 on Build GRCh38
rs902945054 Chr.15: 51456302-51456302 on Build GRCh38
rs760303033 Chr.15: 51456163-51456163 on Build GRCh38

View Full Product Details