Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 85073895-85074136 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOOL
Entrez Gene ID
139322
Gene Name
apolipoprotein O like
Gene Aliases
CXorf33, FAM121A, Mic27, UNQ8193
UniGene
Hs.512181
Gene Chromosome Location
Chr.X: 85003852-85093317 on Build GRCh38
Transcripts
NM_198450.5 XM_011530859.2 XM_017029271.1 XM_017029272.1 XM_017029273.1

SNP Detail

SNP ID Chromosome Location
rs1046490936 Chr.X: 85074111-85074111 on Build GRCh38
rs373265700 Chr.X: 85074013-85074013 on Build GRCh38
rs1000925289 Chr.X: 85074012-85074012 on Build GRCh38
rs750149022 Chr.X: 85074071-85074071 on Build GRCh38
rs762536394 Chr.X: 85073972-85073972 on Build GRCh38
rs933528035 Chr.X: 85073936-85073936 on Build GRCh38
rs761648432 Chr.X: 85074005-85074005 on Build GRCh38
rs865968849 Chr.X: 85074091-85074091 on Build GRCh38
rs761399330 Chr.X: 85074021-85074021 on Build GRCh38
rs34784618 Chr.X: 85074041-85074042 on Build GRCh38
rs138733914 Chr.X: 85074064-85074064 on Build GRCh38
rs753797569 Chr.X: 85074043-85074043 on Build GRCh38
rs766725192 Chr.X: 85074036-85074036 on Build GRCh38
rs768410272 Chr.X: 85073992-85073992 on Build GRCh38
rs961322066 Chr.X: 85074086-85074086 on Build GRCh38
rs750640452 Chr.X: 85074018-85074018 on Build GRCh38
rs778942613 Chr.X: 85074081-85074081 on Build GRCh38
rs1037876555 Chr.X: 85074044-85074044 on Build GRCh38
rs201017526 Chr.X: 85074095-85074095 on Build GRCh38
rs774372566 Chr.X: 85073993-85073993 on Build GRCh38
rs763560828 Chr.X: 85073947-85073947 on Build GRCh38
rs761037702 Chr.X: 85074030-85074030 on Build GRCh38
rs1026666049 Chr.X: 85074112-85074112 on Build GRCh38
rs903817019 Chr.X: 85073998-85073998 on Build GRCh38

View Full Product Details