Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 42756122-42756359 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA6
Entrez Gene ID
8973
Gene Name
cholinergic receptor nicotinic alpha 6 subunit
Gene Aliases
CHNRA6
UniGene
Hs.103128
Gene Chromosome Location
Chr.8: 42752620-42768786 on Build GRCh38
Transcripts
NM_001199279.1 NM_004198.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3367357 c.1033T>G p.F345V Chr.8: 42756166-42756166 on Build GRCh38
1457185 c.916T>C p.Y306H Chr.8: 42756283-42756283 on Build GRCh38
6628737 c.894G>T p.L298L Chr.8: 42756305-42756305 on Build GRCh38
1314068 c.1062G>A p.L354L Chr.8: 42756137-42756137 on Build GRCh38
4519687 c.1023G>A p.V341V Chr.8: 42756176-42756176 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs185610062 Chr.8: 42756224-42756224 on Build GRCh38
rs200829161 Chr.8: 42756268-42756268 on Build GRCh38
rs193137069 Chr.8: 42756142-42756142 on Build GRCh38
rs773917226 Chr.8: 42756271-42756271 on Build GRCh38
rs778753253 Chr.8: 42756204-42756204 on Build GRCh38
rs200072442 Chr.8: 42756313-42756313 on Build GRCh38
rs779838446 Chr.8: 42756217-42756217 on Build GRCh38
rs142629147 Chr.8: 42756300-42756300 on Build GRCh38
rs750671647 Chr.8: 42756305-42756305 on Build GRCh38
rs372377533 Chr.8: 42756153-42756153 on Build GRCh38
rs776324742 Chr.8: 42756232-42756232 on Build GRCh38
rs763093183 Chr.8: 42756315-42756315 on Build GRCh38
rs989145751 Chr.8: 42756263-42756263 on Build GRCh38
rs201530242 Chr.8: 42756202-42756202 on Build GRCh38
rs201193214 Chr.8: 42756174-42756174 on Build GRCh38
rs775696869 Chr.8: 42756287-42756287 on Build GRCh38
rs200533962 Chr.8: 42756335-42756335 on Build GRCh38
rs148473463 Chr.8: 42756147-42756147 on Build GRCh38
rs976951248 Chr.8: 42756207-42756207 on Build GRCh38
rs199921540 Chr.8: 42756288-42756288 on Build GRCh38
rs373535041 Chr.8: 42756191-42756191 on Build GRCh38
rs199675892 Chr.8: 42756215-42756215 on Build GRCh38
rs373608967 Chr.8: 42756336-42756336 on Build GRCh38
rs767679720 Chr.8: 42756293-42756293 on Build GRCh38
rs922882584 Chr.8: 42756209-42756209 on Build GRCh38
rs376849555 Chr.8: 42756220-42756220 on Build GRCh38
rs769758264 Chr.8: 42756264-42756266 on Build GRCh38
rs888527245 Chr.8: 42756246-42756246 on Build GRCh38
rs199892210 Chr.8: 42756253-42756253 on Build GRCh38
rs200331504 Chr.8: 42756244-42756244 on Build GRCh38
rs201833648 Chr.8: 42756245-42756245 on Build GRCh38
rs544312612 Chr.8: 42756334-42756334 on Build GRCh38
rs769343905 Chr.8: 42756250-42756250 on Build GRCh38
rs982008922 Chr.8: 42756210-42756210 on Build GRCh38
rs371233399 Chr.8: 42756309-42756309 on Build GRCh38
rs763481468 Chr.8: 42756186-42756186 on Build GRCh38
rs577047625 Chr.8: 42756143-42756143 on Build GRCh38
rs201852908 Chr.8: 42756280-42756280 on Build GRCh38
rs760634963 Chr.8: 42756289-42756289 on Build GRCh38
rs745799927 Chr.8: 42756211-42756211 on Build GRCh38
rs758246174 Chr.8: 42756213-42756213 on Build GRCh38
rs372414818 Chr.8: 42756275-42756275 on Build GRCh38
rs759926957 Chr.8: 42756281-42756281 on Build GRCh38
rs375238294 Chr.8: 42756302-42756302 on Build GRCh38
rs200513077 Chr.8: 42756198-42756198 on Build GRCh38
rs746856231 Chr.8: 42756323-42756323 on Build GRCh38
rs200381658 Chr.8: 42756149-42756149 on Build GRCh38
rs894446733 Chr.8: 42756327-42756327 on Build GRCh38
rs753856041 Chr.8: 42756312-42756312 on Build GRCh38
rs775611153 Chr.8: 42756197-42756197 on Build GRCh38
rs35812517 Chr.8: 42756258-42756258 on Build GRCh38
rs199691486 Chr.8: 42756193-42756193 on Build GRCh38
rs201044590 Chr.8: 42756299-42756299 on Build GRCh38
rs758336371 Chr.8: 42756320-42756320 on Build GRCh38
rs202140850 Chr.8: 42756308-42756308 on Build GRCh38

View Full Product Details