Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96129997-96130228 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASTL
Entrez Gene ID
431705
Gene Name
astacin-like metallo-endopeptidase (M12 family)
Gene Aliases
SAS1B
UniGene
Hs.447993
Gene Chromosome Location
Chr.2: 96122817-96143059 on Build GRCh38
Transcripts
NM_001002036.3 XM_011511205.2 XM_011511206.2 XM_011511207.2 XM_011511208.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
400576 c.672C>A p.S224R Chr.2: 96130111-96130111 on Build GRCh38
4557172 c.717G>A p.G239G Chr.2: 96130066-96130066 on Build GRCh38
722855 c.659A>T p.K220M Chr.2: 96130124-96130124 on Build GRCh38
6606116 c.684G>A p.T228T Chr.2: 96130099-96130099 on Build GRCh38
4096346 c.703G>A p.V235M Chr.2: 96130080-96130080 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774372393 Chr.2: 96130185-96130185 on Build GRCh38
rs762938662 Chr.2: 96130070-96130070 on Build GRCh38
rs369137669 Chr.2: 96130055-96130055 on Build GRCh38
rs201932296 Chr.2: 96130074-96130074 on Build GRCh38
rs769335040 Chr.2: 96130058-96130058 on Build GRCh38
rs752809649 Chr.2: 96130128-96130128 on Build GRCh38
rs879634697 Chr.2: 96130155-96130155 on Build GRCh38
rs748701062 Chr.2: 96130022-96130022 on Build GRCh38
rs911444546 Chr.2: 96130073-96130073 on Build GRCh38
rs764090414 Chr.2: 96130090-96130090 on Build GRCh38
rs141311153 Chr.2: 96130066-96130066 on Build GRCh38
rs761686236 Chr.2: 96130099-96130099 on Build GRCh38
rs1025406269 Chr.2: 96130038-96130038 on Build GRCh38
rs771132755 Chr.2: 96130040-96130040 on Build GRCh38
rs776358748 Chr.2: 96130051-96130051 on Build GRCh38
rs979594918 Chr.2: 96130117-96130117 on Build GRCh38
rs758892466 Chr.2: 96130137-96130137 on Build GRCh38
rs1028124334 Chr.2: 96130201-96130201 on Build GRCh38
rs201097137 Chr.2: 96130163-96130163 on Build GRCh38
rs757663123 Chr.2: 96130156-96130156 on Build GRCh38
rs760230433 Chr.2: 96130082-96130084 on Build GRCh38
rs767381135 Chr.2: 96130194-96130194 on Build GRCh38
rs778347060 Chr.2: 96130152-96130152 on Build GRCh38
rs199676162 Chr.2: 96130046-96130046 on Build GRCh38
rs572376409 Chr.2: 96130175-96130175 on Build GRCh38
rs767172858 Chr.2: 96130100-96130100 on Build GRCh38
rs369314456 Chr.2: 96130181-96130181 on Build GRCh38
rs1052922960 Chr.2: 96130133-96130133 on Build GRCh38
rs749457 Chr.2: 96130205-96130205 on Build GRCh38
rs774970522 Chr.2: 96130041-96130043 on Build GRCh38
rs774093278 Chr.2: 96130094-96130094 on Build GRCh38
rs747416533 Chr.2: 96130153-96130153 on Build GRCh38
rs1008291244 Chr.2: 96130207-96130207 on Build GRCh38
rs745570881 Chr.2: 96130164-96130164 on Build GRCh38
rs556646524 Chr.2: 96130029-96130029 on Build GRCh38
rs767270648 Chr.2: 96130028-96130039 on Build GRCh38
rs549889647 Chr.2: 96130187-96130187 on Build GRCh38
rs374766318 Chr.2: 96130018-96130018 on Build GRCh38
rs879250506 Chr.2: 96130054-96130054 on Build GRCh38
rs373044804 Chr.2: 96130188-96130188 on Build GRCh38
rs749458 Chr.2: 96130118-96130118 on Build GRCh38
rs373134495 Chr.2: 96130119-96130119 on Build GRCh38
rs775050116 Chr.2: 96130179-96130179 on Build GRCh38
rs767997079 Chr.2: 96130184-96130184 on Build GRCh38
rs754140470 Chr.2: 96130115-96130115 on Build GRCh38
rs747393691 Chr.2: 96130032-96130032 on Build GRCh38

View Full Product Details