Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 119105967-119106226 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COLEC10
Entrez Gene ID
10584
Gene Name
collectin subfamily member 10
Gene Aliases
CL-34, CLL1
UniGene
Hs.176615
Gene Chromosome Location
Chr.8: 118952167-119108455 on Build GRCh38
Transcripts
NM_001324095.1 NM_006438.4 XM_005250756.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1700564 c.649G>A p.E217K Chr.8: 119106006-119106006 on Build GRCh38
3896380 c.634A>T p.N212Y Chr.8: 119105991-119105991 on Build GRCh38
4905036 c.648G>A p.R216R Chr.8: 119106005-119106005 on Build GRCh38
243811 c.652G>A p.G218R Chr.8: 119106009-119106009 on Build GRCh38
6891340 c.811G>A p.E271K Chr.8: 119106168-119106168 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748324624 Chr.8: 119106044-119106044 on Build GRCh38
rs758740979 Chr.8: 119106154-119106154 on Build GRCh38
rs369373077 Chr.8: 119106171-119106171 on Build GRCh38
rs373495591 Chr.8: 119106135-119106135 on Build GRCh38
rs767280625 Chr.8: 119105995-119105995 on Build GRCh38
rs751195682 Chr.8: 119105988-119105988 on Build GRCh38
rs781726945 Chr.8: 119106174-119106174 on Build GRCh38
rs772595750 Chr.8: 119106045-119106045 on Build GRCh38
rs778150778 Chr.8: 119106046-119106046 on Build GRCh38
rs1001345119 Chr.8: 119106004-119106004 on Build GRCh38
rs762273251 Chr.8: 119106183-119106183 on Build GRCh38
rs755122786 Chr.8: 119106116-119106116 on Build GRCh38
rs752989802 Chr.8: 119106145-119106145 on Build GRCh38
rs755106362 Chr.8: 119106030-119106030 on Build GRCh38
rs1039949046 Chr.8: 119106155-119106155 on Build GRCh38
rs1034134375 Chr.8: 119106022-119106022 on Build GRCh38
rs140080059 Chr.8: 119106176-119106176 on Build GRCh38
rs531674603 Chr.8: 119106100-119106100 on Build GRCh38
rs761419555 Chr.8: 119106077-119106077 on Build GRCh38
rs756033494 Chr.8: 119106018-119106018 on Build GRCh38
rs549768640 Chr.8: 119106108-119106108 on Build GRCh38
rs142094873 Chr.8: 119106065-119106065 on Build GRCh38
rs747470853 Chr.8: 119106161-119106161 on Build GRCh38
rs779836343 Chr.8: 119106023-119106023 on Build GRCh38
rs151148307 Chr.8: 119106071-119106071 on Build GRCh38
rs139869623 Chr.8: 119106014-119106014 on Build GRCh38
rs369226811 Chr.8: 119106072-119106072 on Build GRCh38
rs762415988 Chr.8: 119105987-119105987 on Build GRCh38
rs760357008 Chr.8: 119106084-119106084 on Build GRCh38
rs148434952 Chr.8: 119106182-119106182 on Build GRCh38
rs888925405 Chr.8: 119106142-119106142 on Build GRCh38
rs774000075 Chr.8: 119106074-119106074 on Build GRCh38
rs774743364 Chr.8: 119106161-119106164 on Build GRCh38
rs779336705 Chr.8: 119106032-119106032 on Build GRCh38
rs374827000 Chr.8: 119106029-119106029 on Build GRCh38
rs986813847 Chr.8: 119106104-119106104 on Build GRCh38
rs746304259 Chr.8: 119106191-119106191 on Build GRCh38
rs866748212 Chr.8: 119106090-119106090 on Build GRCh38
rs376866289 Chr.8: 119106137-119106137 on Build GRCh38
rs567148567 Chr.8: 119106081-119106081 on Build GRCh38

View Full Product Details