Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 103004642-103004867 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYLC2
Entrez Gene ID
1539
Gene Name
cylicin 2
Gene Aliases
-
UniGene
Hs.3232
Gene Chromosome Location
Chr.9: 102995311-103018488 on Build GRCh38
Transcripts
NM_001340.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6638949 c.247A>G p.R83G Chr.9: 103004761-103004761 on Build GRCh38
7428252 c.287G>A p.R96K Chr.9: 103004801-103004801 on Build GRCh38
4537987 c.248G>C p.R83T Chr.9: 103004762-103004762 on Build GRCh38
6638953 c.236G>A p.R79H Chr.9: 103004750-103004750 on Build GRCh38
752224 c.321G>T p.V107V Chr.9: 103004835-103004835 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746867819 Chr.9: 103004754-103004754 on Build GRCh38
rs569740888 Chr.9: 103004722-103004722 on Build GRCh38
rs141282812 Chr.9: 103004772-103004772 on Build GRCh38
rs761134343 Chr.9: 103004799-103004799 on Build GRCh38
rs767771773 Chr.9: 103004702-103004702 on Build GRCh38
rs753247753 Chr.9: 103004725-103004725 on Build GRCh38
rs113381774 Chr.9: 103004726-103004726 on Build GRCh38
rs143843968 Chr.9: 103004827-103004827 on Build GRCh38
rs967726738 Chr.9: 103004760-103004760 on Build GRCh38
rs112904919 Chr.9: 103004788-103004788 on Build GRCh38
rs775014958 Chr.9: 103004671-103004671 on Build GRCh38
rs146582312 Chr.9: 103004750-103004750 on Build GRCh38
rs375836898 Chr.9: 103004821-103004821 on Build GRCh38
rs1009137314 Chr.9: 103004777-103004777 on Build GRCh38
rs760109206 Chr.9: 103004776-103004776 on Build GRCh38
rs761244512 Chr.9: 103004816-103004816 on Build GRCh38
rs188765977 Chr.9: 103004735-103004735 on Build GRCh38
rs369036643 Chr.9: 103004830-103004830 on Build GRCh38
rs769872615 Chr.9: 103004756-103004756 on Build GRCh38
rs774642858 Chr.9: 103004773-103004773 on Build GRCh38
rs972184895 Chr.9: 103004800-103004800 on Build GRCh38
rs759269481 Chr.9: 103004711-103004711 on Build GRCh38
rs145110008 Chr.9: 103004792-103004792 on Build GRCh38
rs753164725 Chr.9: 103004703-103004703 on Build GRCh38
rs915903047 Chr.9: 103004826-103004826 on Build GRCh38
rs750364071 Chr.9: 103004738-103004738 on Build GRCh38
rs71511703 Chr.9: 103004709-103004709 on Build GRCh38
rs749519557 Chr.9: 103004834-103004834 on Build GRCh38
rs778083431 Chr.9: 103004761-103004761 on Build GRCh38
rs371955196 Chr.9: 103004778-103004778 on Build GRCh38
rs139628328 Chr.9: 103004809-103004809 on Build GRCh38
rs112545316 Chr.9: 103004806-103004806 on Build GRCh38
rs138926365 Chr.9: 103004796-103004796 on Build GRCh38
rs751268360 Chr.9: 103004823-103004823 on Build GRCh38
rs374978801 Chr.9: 103004822-103004822 on Build GRCh38
rs766100136 Chr.9: 103004740-103004740 on Build GRCh38
rs1020142422 Chr.9: 103004759-103004759 on Build GRCh38
rs369157608 Chr.9: 103004699-103004699 on Build GRCh38
rs758395916 Chr.9: 103004748-103004748 on Build GRCh38
rs113473315 Chr.9: 103004801-103004801 on Build GRCh38
rs753500877 Chr.9: 103004814-103004814 on Build GRCh38

View Full Product Details