Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49642308-49642581 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BSN
Entrez Gene ID
8927
Gene Name
bassoon presynaptic cytomatrix protein
Gene Aliases
ZNF231
UniGene
Hs.194684
Gene Chromosome Location
Chr.3: 49554489-49671549 on Build GRCh38
Transcripts
NM_003458.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4584462 c.770C>T p.P257L Chr.3: 49642404-49642404 on Build GRCh38
4507189 c.739C>T p.L247L Chr.3: 49642373-49642373 on Build GRCh38
5885788 c.804_805delCC p.P269fs*11 Chr.3: 49642438-49642439 on Build GRCh38
4513343 c.934C>T p.P312S Chr.3: 49642568-49642568 on Build GRCh38
6613215 c.903G>A p.R301R Chr.3: 49642537-49642537 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868782180 Chr.3: 49642396-49642396 on Build GRCh38
rs776193911 Chr.3: 49642448-49642448 on Build GRCh38
rs749383999 Chr.3: 49642398-49642398 on Build GRCh38
rs763726090 Chr.3: 49642360-49642360 on Build GRCh38
rs192482729 Chr.3: 49642354-49642354 on Build GRCh38
rs780401178 Chr.3: 49642563-49642563 on Build GRCh38
rs979436660 Chr.3: 49642464-49642464 on Build GRCh38
rs772010793 Chr.3: 49642428-49642428 on Build GRCh38
rs1002436540 Chr.3: 49642537-49642537 on Build GRCh38
rs773529145 Chr.3: 49642361-49642361 on Build GRCh38
rs1001920400 Chr.3: 49642369-49642369 on Build GRCh38
rs753923690 Chr.3: 49642506-49642506 on Build GRCh38
rs761300724 Chr.3: 49642463-49642463 on Build GRCh38
rs750415602 Chr.3: 49642538-49642538 on Build GRCh38
rs968160275 Chr.3: 49642337-49642337 on Build GRCh38
rs775327878 Chr.3: 49642334-49642334 on Build GRCh38
rs934089830 Chr.3: 49642488-49642488 on Build GRCh38
rs942871517 Chr.3: 49642476-49642476 on Build GRCh38
rs564597250 Chr.3: 49642392-49642392 on Build GRCh38
rs764287938 Chr.3: 49642502-49642502 on Build GRCh38
rs1023304604 Chr.3: 49642330-49642330 on Build GRCh38
rs757329210 Chr.3: 49642512-49642512 on Build GRCh38
rs745649866 Chr.3: 49642406-49642406 on Build GRCh38
rs751952854 Chr.3: 49642384-49642384 on Build GRCh38
rs768227077 Chr.3: 49642441-49642441 on Build GRCh38
rs775241611 Chr.3: 49642433-49642433 on Build GRCh38
rs200211104 Chr.3: 49642496-49642496 on Build GRCh38
rs146673139 Chr.3: 49642529-49642529 on Build GRCh38
rs757213353 Chr.3: 49642404-49642404 on Build GRCh38
rs746661090 Chr.3: 49642434-49642434 on Build GRCh38
rs755377052 Chr.3: 49642385-49642385 on Build GRCh38
rs115552063 Chr.3: 49642393-49642393 on Build GRCh38
rs149931635 Chr.3: 49642338-49642338 on Build GRCh38
rs758197685 Chr.3: 49642551-49642551 on Build GRCh38
rs926104821 Chr.3: 49642469-49642469 on Build GRCh38
rs200890584 Chr.3: 49642497-49642497 on Build GRCh38
rs771441917 Chr.3: 49642487-49642487 on Build GRCh38
rs763532940 Chr.3: 49642366-49642366 on Build GRCh38
rs758826087 Chr.3: 49642504-49642504 on Build GRCh38
rs370650979 Chr.3: 49642498-49642498 on Build GRCh38
rs923720886 Chr.3: 49642363-49642363 on Build GRCh38
rs766858181 Chr.3: 49642372-49642372 on Build GRCh38
rs931331292 Chr.3: 49642439-49642439 on Build GRCh38
rs544923830 Chr.3: 49642389-49642389 on Build GRCh38
rs760872406 Chr.3: 49642499-49642499 on Build GRCh38
rs1047157198 Chr.3: 49642455-49642455 on Build GRCh38
rs202140904 Chr.3: 49642493-49642493 on Build GRCh38

View Full Product Details