Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 24361246-24361507 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNG3
Entrez Gene ID
10368
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 3
Gene Aliases
-
UniGene
Hs.7235
Gene Chromosome Location
Chr.16: 24255553-24362416 on Build GRCh38
Transcripts
NM_006539.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3507984 c.489C>T p.D163D Chr.16: 24361404-24361404 on Build GRCh38
256526 c.568G>T p.E190* Chr.16: 24361483-24361483 on Build GRCh38
7308880 c.576A>G p.V192V Chr.16: 24361491-24361491 on Build GRCh38
5496275 c.554C>G p.S185C Chr.16: 24361469-24361469 on Build GRCh38
6144038 c.437-2A>T p.? Chr.16: 24361350-24361350 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745589919 Chr.16: 24361339-24361339 on Build GRCh38
rs748855302 Chr.16: 24361333-24361333 on Build GRCh38
rs899173012 Chr.16: 24361299-24361299 on Build GRCh38
rs141984716 Chr.16: 24361273-24361273 on Build GRCh38
rs371160374 Chr.16: 24361398-24361398 on Build GRCh38
rs927880920 Chr.16: 24361275-24361275 on Build GRCh38
rs752379957 Chr.16: 24361326-24361326 on Build GRCh38
rs865859398 Chr.16: 24361473-24361473 on Build GRCh38
rs755805828 Chr.16: 24361327-24361327 on Build GRCh38
rs750567092 Chr.16: 24361313-24361313 on Build GRCh38
rs201012526 Chr.16: 24361458-24361458 on Build GRCh38
rs762094078 Chr.16: 24361383-24361383 on Build GRCh38
rs867945636 Chr.16: 24361448-24361448 on Build GRCh38
rs763168467 Chr.16: 24361312-24361313 on Build GRCh38
rs761934623 Chr.16: 24361309-24361309 on Build GRCh38
rs770185514 Chr.16: 24361395-24361395 on Build GRCh38
rs766388248 Chr.16: 24361328-24361329 on Build GRCh38
rs988054547 Chr.16: 24361312-24361312 on Build GRCh38
rs867799411 Chr.16: 24361479-24361479 on Build GRCh38
rs946560806 Chr.16: 24361368-24361368 on Build GRCh38
rs755386590 Chr.16: 24361321-24361321 on Build GRCh38
rs776950399 Chr.16: 24361370-24361370 on Build GRCh38
rs765436999 Chr.16: 24361310-24361310 on Build GRCh38
rs767894450 Chr.16: 24361449-24361449 on Build GRCh38
rs753277538 Chr.16: 24361474-24361474 on Build GRCh38
rs766980359 Chr.16: 24361317-24361317 on Build GRCh38
rs200040220 Chr.16: 24361452-24361452 on Build GRCh38
rs988067948 Chr.16: 24361467-24361467 on Build GRCh38
rs771922151 Chr.16: 24361342-24361342 on Build GRCh38
rs868528231 Chr.16: 24361352-24361352 on Build GRCh38
rs780662089 Chr.16: 24361324-24361324 on Build GRCh38
rs1034130875 Chr.16: 24361288-24361288 on Build GRCh38
rs150191653 Chr.16: 24361415-24361415 on Build GRCh38
rs145243165 Chr.16: 24361314-24361314 on Build GRCh38
rs766742794 Chr.16: 24361399-24361399 on Build GRCh38
rs370738159 Chr.16: 24361334-24361334 on Build GRCh38
rs777243604 Chr.16: 24361328-24361328 on Build GRCh38
rs950931829 Chr.16: 24361410-24361410 on Build GRCh38
rs140935639 Chr.16: 24361396-24361396 on Build GRCh38
rs779898573 Chr.16: 24361477-24361477 on Build GRCh38
rs531052899 Chr.16: 24361407-24361407 on Build GRCh38

View Full Product Details