Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 47258336-47258609 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC9
Entrez Gene ID
26093
Gene Name
coiled-coil domain containing 9
Gene Aliases
-
UniGene
Hs.227782
Gene Chromosome Location
Chr.19: 47256474-47271958 on Build GRCh38
Transcripts
NM_015603.2 XM_017026578.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6293809 c.4-8G>A p.? Chr.19: 47258551-47258551 on Build GRCh38
1612471 c.4-7G>T p.? Chr.19: 47258552-47258552 on Build GRCh38
6257570 c.51G>T p.L17F Chr.19: 47258606-47258606 on Build GRCh38
1394876 c.16G>A p.D6N Chr.19: 47258571-47258571 on Build GRCh38
5744001 c.3+2T>C p.? Chr.19: 47258405-47258405 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758408601 Chr.19: 47258450-47258450 on Build GRCh38
rs777386353 Chr.19: 47258582-47258585 on Build GRCh38
rs370434658 Chr.19: 47258366-47258366 on Build GRCh38
rs756373331 Chr.19: 47258584-47258586 on Build GRCh38
rs114448157 Chr.19: 47258474-47258474 on Build GRCh38
rs767409626 Chr.19: 47258521-47258522 on Build GRCh38
rs758044756 Chr.19: 47258433-47258434 on Build GRCh38
rs749114613 Chr.19: 47258398-47258398 on Build GRCh38
rs368143959 Chr.19: 47258395-47258395 on Build GRCh38
rs752506366 Chr.19: 47258411-47258411 on Build GRCh38
rs750171411 Chr.19: 47258550-47258553 on Build GRCh38
rs1040575970 Chr.19: 47258482-47258482 on Build GRCh38
rs748498221 Chr.19: 47258386-47258386 on Build GRCh38
rs979307923 Chr.19: 47258425-47258425 on Build GRCh38
rs764009874 Chr.19: 47258446-47258446 on Build GRCh38
rs375789072 Chr.19: 47258358-47258358 on Build GRCh38
rs902349516 Chr.19: 47258583-47258583 on Build GRCh38
rs202021250 Chr.19: 47258551-47258551 on Build GRCh38
rs368291098 Chr.19: 47258572-47258572 on Build GRCh38
rs762493015 Chr.19: 47258394-47258394 on Build GRCh38
rs768865729 Chr.19: 47258377-47258377 on Build GRCh38
rs940279516 Chr.19: 47258501-47258501 on Build GRCh38
rs113328209 Chr.19: 47258545-47258545 on Build GRCh38
rs990535842 Chr.19: 47258457-47258457 on Build GRCh38
rs950281313 Chr.19: 47258472-47258472 on Build GRCh38
rs200133167 Chr.19: 47258526-47258526 on Build GRCh38
rs759009445 Chr.19: 47258533-47258533 on Build GRCh38
rs770326061 Chr.19: 47258388-47258388 on Build GRCh38
rs755066857 Chr.19: 47258581-47258581 on Build GRCh38
rs755192830 Chr.19: 47258518-47258518 on Build GRCh38
rs12985905 Chr.19: 47258389-47258389 on Build GRCh38
rs539655267 Chr.19: 47258571-47258571 on Build GRCh38
rs767195152 Chr.19: 47258535-47258535 on Build GRCh38
rs770295777 Chr.19: 47258521-47258521 on Build GRCh38
rs1022650183 Chr.19: 47258555-47258555 on Build GRCh38
rs748527842 Chr.19: 47258519-47258519 on Build GRCh38
rs970017179 Chr.19: 47258570-47258570 on Build GRCh38
rs557910476 Chr.19: 47258549-47258549 on Build GRCh38
rs889007681 Chr.19: 47258421-47258421 on Build GRCh38
rs1020817713 Chr.19: 47258576-47258576 on Build GRCh38
rs1038630934 Chr.19: 47258507-47258507 on Build GRCh38
rs1027440591 Chr.19: 47258523-47258523 on Build GRCh38
rs774104820 Chr.19: 47258396-47258396 on Build GRCh38
rs773788277 Chr.19: 47258522-47258522 on Build GRCh38
rs775063400 Chr.19: 47258542-47258542 on Build GRCh38
rs767125951 Chr.19: 47258402-47258402 on Build GRCh38
rs137964332 Chr.19: 47258567-47258567 on Build GRCh38
rs372031704 Chr.19: 47258515-47258515 on Build GRCh38
rs763885165 Chr.19: 47258550-47258550 on Build GRCh38
rs757211911 Chr.19: 47258559-47258559 on Build GRCh38
rs374782034 Chr.19: 47258375-47258375 on Build GRCh38
rs773891745 Chr.19: 47258527-47258527 on Build GRCh38
rs1032005161 Chr.19: 47258426-47258426 on Build GRCh38
rs753790393 Chr.19: 47258447-47258447 on Build GRCh38
rs777028907 Chr.19: 47258383-47258383 on Build GRCh38
rs917633839 Chr.19: 47258466-47258466 on Build GRCh38
rs759256720 Chr.19: 47258400-47258400 on Build GRCh38
rs755929814 Chr.19: 47258414-47258414 on Build GRCh38
rs887429019 Chr.19: 47258546-47258546 on Build GRCh38

View Full Product Details