Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 152302632-152302879 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLG
Entrez Gene ID
2312
Gene Name
filaggrin
Gene Aliases
ATOD2
UniGene
Hs.654510
Gene Chromosome Location
Chr.1: 152302175-152325203 on Build GRCh38
Transcripts
NM_002016.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3473947 c.12167G>A p.R4056K Chr.1: 152302719-152302719 on Build GRCh38
5605726 c.12008C>T p.S4003F Chr.1: 152302878-152302878 on Build GRCh38
6664554 c.12019G>C p.V4007L Chr.1: 152302867-152302867 on Build GRCh38
2184898 c.12070C>T p.H4024Y Chr.1: 152302816-152302816 on Build GRCh38
2184897 c.12072T>A p.H4024Q Chr.1: 152302814-152302814 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746918389 Chr.1: 152302675-152302675 on Build GRCh38
rs369153121 Chr.1: 152302766-152302766 on Build GRCh38
rs926016191 Chr.1: 152302744-152302744 on Build GRCh38
rs758681801 Chr.1: 152302706-152302711 on Build GRCh38
rs1033493107 Chr.1: 152302772-152302772 on Build GRCh38
rs146466242 Chr.1: 152302822-152302822 on Build GRCh38
rs760828124 Chr.1: 152302750-152302750 on Build GRCh38
rs746954478 Chr.1: 152302741-152302741 on Build GRCh38
rs760032456 Chr.1: 152302739-152302739 on Build GRCh38
rs367608187 Chr.1: 152302663-152302663 on Build GRCh38
rs561151352 Chr.1: 152302802-152302802 on Build GRCh38
rs751610435 Chr.1: 152302779-152302779 on Build GRCh38
rs1053086412 Chr.1: 152302767-152302767 on Build GRCh38
rs773484179 Chr.1: 152302712-152302712 on Build GRCh38
rs1039173304 Chr.1: 152302737-152302737 on Build GRCh38
rs771179731 Chr.1: 152302729-152302729 on Build GRCh38
rs768335641 Chr.1: 152302689-152302689 on Build GRCh38
rs184470276 Chr.1: 152302754-152302754 on Build GRCh38
rs372573825 Chr.1: 152302830-152302830 on Build GRCh38
rs1002017153 Chr.1: 152302723-152302723 on Build GRCh38
rs761932378 Chr.1: 152302834-152302834 on Build GRCh38
rs752974777 Chr.1: 152302660-152302660 on Build GRCh38
rs763418475 Chr.1: 152302715-152302715 on Build GRCh38
rs769442034 Chr.1: 152302705-152302705 on Build GRCh38
rs150630644 Chr.1: 152302748-152302748 on Build GRCh38
rs752715330 Chr.1: 152302749-152302749 on Build GRCh38
rs780392363 Chr.1: 152302792-152302792 on Build GRCh38
rs371451436 Chr.1: 152302677-152302677 on Build GRCh38
rs1046661358 Chr.1: 152302803-152302803 on Build GRCh38
rs750183118 Chr.1: 152302666-152302666 on Build GRCh38
rs144304274 Chr.1: 152302753-152302753 on Build GRCh38
rs536917379 Chr.1: 152302668-152302668 on Build GRCh38
rs117440780 Chr.1: 152302796-152302796 on Build GRCh38
rs552696097 Chr.1: 152302818-152302818 on Build GRCh38
rs988056657 Chr.1: 152302695-152302695 on Build GRCh38
rs774631324 Chr.1: 152302801-152302801 on Build GRCh38
rs779978288 Chr.1: 152302778-152302778 on Build GRCh38
rs148776256 Chr.1: 152302781-152302781 on Build GRCh38
rs766276866 Chr.1: 152302842-152302842 on Build GRCh38
rs919533300 Chr.1: 152302688-152302688 on Build GRCh38
rs747657151 Chr.1: 152302787-152302787 on Build GRCh38
rs751354626 Chr.1: 152302852-152302852 on Build GRCh38
rs774898602 Chr.1: 152302733-152302733 on Build GRCh38
rs759542688 Chr.1: 152302710-152302710 on Build GRCh38
rs776967180 Chr.1: 152302832-152302832 on Build GRCh38
rs139645876 Chr.1: 152302701-152302701 on Build GRCh38
rs150330138 Chr.1: 152302800-152302800 on Build GRCh38
rs764336258 Chr.1: 152302752-152302752 on Build GRCh38
rs780178855 Chr.1: 152302671-152302671 on Build GRCh38
rs370295034 Chr.1: 152302829-152302829 on Build GRCh38
rs943449961 Chr.1: 152302703-152302703 on Build GRCh38
rs754909247 Chr.1: 152302780-152302780 on Build GRCh38
rs76330665 Chr.1: 152302797-152302797 on Build GRCh38
rs760606297 Chr.1: 152302664-152302664 on Build GRCh38

View Full Product Details