Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 92050372-92050633 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHX4
Entrez Gene ID
253152
Gene Name
epoxide hydrolase 4
Gene Aliases
ABHD7, EH4, EPHXRP
UniGene
Hs.201555
Gene Chromosome Location
Chr.1: 92029976-92063536 on Build GRCh38
Transcripts
NM_173567.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5596661 c.668C>T p.P223L Chr.1: 92050380-92050380 on Build GRCh38
3735365 c.672G>C p.W224C Chr.1: 92050384-92050384 on Build GRCh38
6127858 c.696A>T p.I232I Chr.1: 92050408-92050408 on Build GRCh38
4556727 c.700G>A p.D234N Chr.1: 92050412-92050412 on Build GRCh38
4144251 c.705C>A p.F235L Chr.1: 92050417-92050417 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143152717 Chr.1: 92050489-92050489 on Build GRCh38
rs753233915 Chr.1: 92050447-92050447 on Build GRCh38
rs561482527 Chr.1: 92050582-92050582 on Build GRCh38
rs780190999 Chr.1: 92050449-92050451 on Build GRCh38
rs747226029 Chr.1: 92050448-92050449 on Build GRCh38
rs114058100 Chr.1: 92050541-92050541 on Build GRCh38
rs754105103 Chr.1: 92050410-92050410 on Build GRCh38
rs901033581 Chr.1: 92050456-92050456 on Build GRCh38
rs766902218 Chr.1: 92050406-92050406 on Build GRCh38
rs553904867 Chr.1: 92050498-92050498 on Build GRCh38
rs541494978 Chr.1: 92050436-92050439 on Build GRCh38
rs1034040127 Chr.1: 92050531-92050531 on Build GRCh38
rs894609167 Chr.1: 92050588-92050588 on Build GRCh38
rs751260926 Chr.1: 92050458-92050458 on Build GRCh38
rs1018512217 Chr.1: 92050476-92050476 on Build GRCh38
rs113987778 Chr.1: 92050514-92050514 on Build GRCh38
rs977424025 Chr.1: 92050530-92050530 on Build GRCh38
rs945998621 Chr.1: 92050516-92050516 on Build GRCh38
rs757970379 Chr.1: 92050454-92050454 on Build GRCh38
rs1025526677 Chr.1: 92050493-92050493 on Build GRCh38
rs183929008 Chr.1: 92050434-92050434 on Build GRCh38
rs781069308 Chr.1: 92050466-92050466 on Build GRCh38
rs777231700 Chr.1: 92050457-92050457 on Build GRCh38
rs760120697 Chr.1: 92050429-92050429 on Build GRCh38

View Full Product Details