Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48298222-48298447 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC114
Entrez Gene ID
93233
Gene Name
coiled-coil domain containing 114
Gene Aliases
CILD20
UniGene
Hs.112645
Gene Chromosome Location
Chr.19: 48296452-48322793 on Build GRCh38
Transcripts
NM_144577.3 XM_005259413.3 XM_005259414.3 XM_005259415.3 XM_005259416.4 XM_011527515.2 XM_011527516.2 XM_011527517.1 XM_017027483.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
380465 c.1162G>T p.D388Y Chr.19: 48298308-48298308 on Build GRCh38
4079822 c.545G>A p.S182N Chr.19: 48298304-48298304 on Build GRCh38
998920 c.1214G>A p.R405Q Chr.19: 48298256-48298256 on Build GRCh38
6004280 c.520C>A p.L174I Chr.19: 48298329-48298329 on Build GRCh38
380466 c.541G>T p.D181Y Chr.19: 48298308-48298308 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751149003 Chr.19: 48298245-48298245 on Build GRCh38
rs752177994 Chr.19: 48298358-48298358 on Build GRCh38
rs924950486 Chr.19: 48298420-48298420 on Build GRCh38
rs769414060 Chr.19: 48298363-48298363 on Build GRCh38
rs267605561 Chr.19: 48298285-48298285 on Build GRCh38
rs1029289808 Chr.19: 48298366-48298366 on Build GRCh38
rs755803759 Chr.19: 48298364-48298364 on Build GRCh38
rs369830434 Chr.19: 48298355-48298355 on Build GRCh38
rs182947158 Chr.19: 48298379-48298379 on Build GRCh38
rs147082322 Chr.19: 48298256-48298256 on Build GRCh38
rs35930441 Chr.19: 48298306-48298306 on Build GRCh38
rs776940748 Chr.19: 48298334-48298334 on Build GRCh38
rs375942259 Chr.19: 48298294-48298294 on Build GRCh38
rs560134827 Chr.19: 48298308-48298308 on Build GRCh38
rs555664154 Chr.19: 48298309-48298309 on Build GRCh38
rs977829767 Chr.19: 48298413-48298413 on Build GRCh38
rs770121868 Chr.19: 48298338-48298338 on Build GRCh38
rs755924787 Chr.19: 48298269-48298269 on Build GRCh38
rs747874704 Chr.19: 48298344-48298344 on Build GRCh38
rs987669658 Chr.19: 48298377-48298377 on Build GRCh38
rs763730140 Chr.19: 48298378-48298378 on Build GRCh38
rs200855924 Chr.19: 48298304-48298304 on Build GRCh38
rs949996465 Chr.19: 48298404-48298404 on Build GRCh38
rs138500585 Chr.19: 48298257-48298257 on Build GRCh38
rs531078118 Chr.19: 48298397-48298397 on Build GRCh38
rs200024308 Chr.19: 48298346-48298346 on Build GRCh38
rs766968324 Chr.19: 48298345-48298345 on Build GRCh38
rs937146147 Chr.19: 48298374-48298374 on Build GRCh38
rs548920176 Chr.19: 48298353-48298353 on Build GRCh38
rs201757586 Chr.19: 48298268-48298268 on Build GRCh38
rs777592671 Chr.19: 48298289-48298289 on Build GRCh38
rs748335075 Chr.19: 48298337-48298337 on Build GRCh38
rs776150959 Chr.19: 48298270-48298270 on Build GRCh38
rs954925130 Chr.19: 48298368-48298368 on Build GRCh38
rs768549288 Chr.19: 48298313-48298313 on Build GRCh38
rs974997306 Chr.19: 48298357-48298357 on Build GRCh38

View Full Product Details