Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 85801910-85802171 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CADM2
Entrez Gene ID
253559
Gene Name
cell adhesion molecule 2
Gene Aliases
IGSF4D, NECL3, Necl-3, SynCAM 2, synCAM2
UniGene
Hs.164578
Gene Chromosome Location
Chr.3: 84958982-86074429 on Build GRCh38
Transcripts
NM_001167674.1 NM_001167675.1 NM_001256502.1 NM_001256503.1 NM_001256504.1 NM_001256505.1 NM_153184.3 XM_006713081.3 XM_006713082.3 XM_017006062.1 XM_017006063.1 NR_046752.1

Gene Symbol
CADM2-AS2
Entrez Gene ID
100874037
Gene Name
CADM2 antisense RNA 2
Gene Aliases
-
UniGene
Hs.570566
Gene Chromosome Location
Chr.3: 85799987-85828050 on Build GRCh38
Transcripts
NM_001167674.1 NM_001167675.1 NM_001256502.1 NM_001256503.1 NM_001256504.1 NM_001256505.1 NM_153184.3 XM_006713081.3 XM_006713082.3 XM_017006062.1 XM_017006063.1 NR_046752.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7230824 c.162G>C p.R54S Chr.3: 85802120-85802120 on Build GRCh38
3993331 c.166G>C p.D56H Chr.3: 85802124-85802124 on Build GRCh38
1718310 c.189A>C p.Q63H Chr.3: 85802168-85802168 on Build GRCh38
4984585 c.137G>T p.G46V Chr.3: 85802095-85802095 on Build GRCh38
309823 c.147T>G p.D49E Chr.3: 85802126-85802126 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11713930 Chr.3: 85802048-85802048 on Build GRCh38
rs563233143 Chr.3: 85801957-85801957 on Build GRCh38
rs763742405 Chr.3: 85802069-85802069 on Build GRCh38
rs748270193 Chr.3: 85802023-85802023 on Build GRCh38
rs886177014 Chr.3: 85802020-85802020 on Build GRCh38
rs745768336 Chr.3: 85802141-85802141 on Build GRCh38
rs779261257 Chr.3: 85802119-85802119 on Build GRCh38
rs146842915 Chr.3: 85802120-85802120 on Build GRCh38
rs750915514 Chr.3: 85802085-85802085 on Build GRCh38
rs183174165 Chr.3: 85802044-85802044 on Build GRCh38
rs779067028 Chr.3: 85802014-85802014 on Build GRCh38
rs201769601 Chr.3: 85802025-85802025 on Build GRCh38
rs186227152 Chr.3: 85802084-85802084 on Build GRCh38
rs761375901 Chr.3: 85802075-85802075 on Build GRCh38
rs7613828 Chr.3: 85802096-85802096 on Build GRCh38
rs920414156 Chr.3: 85801967-85801967 on Build GRCh38
rs867219031 Chr.3: 85802064-85802064 on Build GRCh38
rs762553115 Chr.3: 85802060-85802060 on Build GRCh38
rs769609286 Chr.3: 85802040-85802040 on Build GRCh38
rs755190295 Chr.3: 85802111-85802111 on Build GRCh38
rs1047493609 Chr.3: 85802019-85802019 on Build GRCh38
rs991070097 Chr.3: 85801962-85801962 on Build GRCh38
rs191313257 Chr.3: 85801946-85801946 on Build GRCh38
rs547176685 Chr.3: 85802088-85802088 on Build GRCh38
rs759431115 Chr.3: 85802039-85802039 on Build GRCh38
rs769754695 Chr.3: 85802147-85802147 on Build GRCh38
rs751014657 Chr.3: 85802070-85802070 on Build GRCh38
rs141396432 Chr.3: 85802009-85802009 on Build GRCh38
rs772099802 Chr.3: 85802024-85802024 on Build GRCh38

View Full Product Details