Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 53992863-53993094 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNG6
Entrez Gene ID
59285
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 6
Gene Aliases
-
UniGene
Hs.631560
Gene Chromosome Location
Chr.19: 53991149-54012666 on Build GRCh38
Transcripts
NM_031897.2 NM_145814.1 NM_145815.1 NR_102308.1 XM_006723316.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4881411 c.208G>T p.A70S Chr.19: 53993085-53993085 on Build GRCh38
1291011 c.58C>T p.R20W Chr.19: 53992935-53992935 on Build GRCh38
7105245 c.28G>A p.E10K Chr.19: 53992905-53992905 on Build GRCh38
5647633 c.40C>A p.R14R Chr.19: 53992917-53992917 on Build GRCh38
4611775 c.44delG p.A17fs*15 Chr.19: 53992921-53992921 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764163084 Chr.19: 53992937-53992937 on Build GRCh38
rs538013052 Chr.19: 53992917-53992934 on Build GRCh38
rs10424407 Chr.19: 53993075-53993075 on Build GRCh38
rs776514934 Chr.19: 53993037-53993037 on Build GRCh38
rs746712453 Chr.19: 53993002-53993002 on Build GRCh38
rs765284445 Chr.19: 53992914-53992914 on Build GRCh38
rs778695297 Chr.19: 53992929-53992960 on Build GRCh38
rs1036759489 Chr.19: 53992985-53992985 on Build GRCh38
rs975999615 Chr.19: 53992938-53992938 on Build GRCh38
rs776676191 Chr.19: 53992903-53992903 on Build GRCh38
rs769787760 Chr.19: 53993072-53993072 on Build GRCh38
rs747728201 Chr.19: 53992882-53992882 on Build GRCh38
rs571070238 Chr.19: 53992955-53992955 on Build GRCh38
rs963016013 Chr.19: 53993077-53993077 on Build GRCh38
rs377123441 Chr.19: 53992975-53992975 on Build GRCh38
rs759591468 Chr.19: 53992904-53992904 on Build GRCh38
rs745685098 Chr.19: 53993071-53993071 on Build GRCh38
rs909294734 Chr.19: 53992958-53992958 on Build GRCh38
rs780619316 Chr.19: 53992950-53992950 on Build GRCh38
rs756574726 Chr.19: 53992949-53992949 on Build GRCh38
rs751864902 Chr.19: 53992941-53992941 on Build GRCh38
rs770825108 Chr.19: 53993015-53993015 on Build GRCh38
rs775391041 Chr.19: 53992917-53992917 on Build GRCh38
rs746770772 Chr.19: 53992889-53992889 on Build GRCh38
rs201414460 Chr.19: 53992946-53992946 on Build GRCh38
rs904562967 Chr.19: 53992921-53992921 on Build GRCh38
rs575512500 Chr.19: 53992920-53992921 on Build GRCh38
rs897797109 Chr.19: 53993045-53993045 on Build GRCh38
rs567490160 Chr.19: 53992923-53992923 on Build GRCh38
rs970294628 Chr.19: 53993047-53993047 on Build GRCh38
rs767806691 Chr.19: 53992913-53992933 on Build GRCh38
rs771562119 Chr.19: 53992885-53992885 on Build GRCh38
rs534917640 Chr.19: 53992942-53992942 on Build GRCh38
rs762329743 Chr.19: 53992907-53992936 on Build GRCh38
rs984405608 Chr.19: 53993063-53993063 on Build GRCh38
rs113333594 Chr.19: 53993036-53993036 on Build GRCh38
rs770913214 Chr.19: 53992901-53992901 on Build GRCh38
rs750808856 Chr.19: 53992948-53992948 on Build GRCh38
rs763098225 Chr.19: 53992926-53992926 on Build GRCh38

View Full Product Details