Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 54113027-54113288 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL15
Entrez Gene ID
54622
Gene Name
ADP ribosylation factor like GTPase 15
Gene Aliases
ARFRP2
UniGene
Hs.659125
Gene Chromosome Location
Chr.5: 53883942-54310590 on Build GRCh38
Transcripts
NM_019087.2 XM_011543498.2 XM_011543499.2 XM_011543500.2 XM_017009598.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3616986 c.407C>T p.P136L Chr.5: 54113257-54113257 on Build GRCh38
7185442 c.443C>A p.A148E Chr.5: 54113221-54113221 on Build GRCh38
266038 c.381A>G p.I127M Chr.5: 54113247-54113247 on Build GRCh38
3855558 c.366T>C p.T122T Chr.5: 54113262-54113262 on Build GRCh38
7185443 c.407C>A p.A136E Chr.5: 54113221-54113221 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1054253877 Chr.5: 54113239-54113239 on Build GRCh38
rs770235548 Chr.5: 54113199-54113199 on Build GRCh38
rs750906027 Chr.5: 54113121-54113121 on Build GRCh38
rs765953485 Chr.5: 54113225-54113225 on Build GRCh38
rs185392005 Chr.5: 54113105-54113105 on Build GRCh38
rs752467549 Chr.5: 54113246-54113246 on Build GRCh38
rs983118537 Chr.5: 54113114-54113114 on Build GRCh38
rs775876885 Chr.5: 54113204-54113204 on Build GRCh38
rs767150861 Chr.5: 54113214-54113214 on Build GRCh38
rs950037330 Chr.5: 54113154-54113154 on Build GRCh38
rs981361687 Chr.5: 54113158-54113158 on Build GRCh38
rs747914970 Chr.5: 54113164-54113164 on Build GRCh38
rs765097364 Chr.5: 54113237-54113237 on Build GRCh38
rs1002098375 Chr.5: 54113193-54113193 on Build GRCh38
rs1004083915 Chr.5: 54113144-54113144 on Build GRCh38
rs202159658 Chr.5: 54113229-54113229 on Build GRCh38
rs756594407 Chr.5: 54113148-54113148 on Build GRCh38
rs776986619 Chr.5: 54113186-54113186 on Build GRCh38
rs77393256 Chr.5: 54113234-54113234 on Build GRCh38
rs1039362128 Chr.5: 54113080-54113080 on Build GRCh38
rs778149105 Chr.5: 54113179-54113179 on Build GRCh38
rs562884023 Chr.5: 54113216-54113216 on Build GRCh38
rs181100044 Chr.5: 54113104-54113104 on Build GRCh38
rs758338658 Chr.5: 54113265-54113265 on Build GRCh38
rs771197031 Chr.5: 54113185-54113185 on Build GRCh38
rs528597163 Chr.5: 54113157-54113157 on Build GRCh38
rs746428279 Chr.5: 54113192-54113192 on Build GRCh38
rs758632862 Chr.5: 54113168-54113168 on Build GRCh38
rs772906106 Chr.5: 54113215-54113215 on Build GRCh38
rs563684982 Chr.5: 54113097-54113097 on Build GRCh38
rs564580809 Chr.5: 54113124-54113124 on Build GRCh38
rs747084197 Chr.5: 54113180-54113180 on Build GRCh38
rs1025195020 Chr.5: 54113147-54113147 on Build GRCh38
rs542120721 Chr.5: 54113077-54113077 on Build GRCh38
rs763322358 Chr.5: 54113210-54113210 on Build GRCh38

View Full Product Details